Skip to Content
Merck
All Photos(2)

Key Documents

MABC95

Sigma-Aldrich

Anti-S1PR2/EDG5 Antibody, clone 6E8.1

clone 6E8.1, from mouse

Synonym(s):

Sphingosine 1-phosphate receptor 2, S1P receptor 2, S1P2, Endothelial differentiation G-protein coupled receptor 5, Sphingosine 1-phosphate receptor Edg-5, S1P receptor Edg-5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6E8.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... S1PR2(9294)

General description

Sphingosine 1-phosphate receptor 2 (S1PR2), otherwise known as EDG5, is a ubiquitous G-protein coupled receptor for lysosphingolipid sphingosine 1-phosphate (S1P). It is coupled to Gi, G12/13, and Gq pathways to regulate a number of second messengers and effector molecules, including cAMP, PLC, MAPK, RhoA, and Rac. S1PR2 mediates the effects of S1P on cell motility and embryonic development, among other cellular processes.

Specificity

This antibody recognizes the cytoplasmic domain of S1PR2/EDG5.

Immunogen

GST-tagged recombinant protein corresponding to the cytoplasmic domain of human S1PR2/EDG5.

Application

Anti-S1PR2/EDG5 Antibody, clone 6E8.1 detects level of S1PR2/EDG5 & has been published & validated for use in Western Blotting, IHC(P).
Immunohistochemistry Analysis: A representative lot detected S1PR2/EDG5 in human tonsil, liver, colorectal cancer tissues.

Quality

Evaluated by Western Blot in human liver tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected S1PR2/EDG5 in 10 mg of human liver tissue lysate.

Target description

~35 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T H Beckham et al.
Oncogenesis, 2, e49-e49 (2013-06-05)
Acid ceramidase (AC) is overexpressed in most prostate tumors and confers oncogenic phenotypes to prostate cancer cells. AC modulates the cellular balance between ceramide, sphingosine and sphingosine 1-phosphate (S1P). These bioactive sphingolipids have diverse, powerful and often oppositional impacts on
Xiao-Jing Sun et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 32(8), 1313-1322 (2017-02-17)
C5a plays a crucial role in anti-neutrophil cytoplasmic antibody (ANCA)-mediated neutrophil recruitment and activation. Our previous studies found that the interaction between sphingosine-1-phosphate (S1P) and C5a plays an important role in the ANCA-mediated activation of neutrophils. In the current study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service