MAB1393-I
Anti-PECAM-1 (CD31) Antibody, clone TLD-3A12
clone TLD-3A12, from mouse
Synonym(s):
Platelet endothelial cell adhesion molecule, CD31
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
TLD-3A12, monoclonal
species reactivity
human, rat
technique(s)
ELISA: suitable
flow cytometry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... PECAM1(5175)
rat ... Pecam1(29583)
General description
Specificity
Immunogen
Application
Flow Cytometry Analysis: 1 µg from a representative lot detected PECAM-1 (CD31) in one million rat splenocytes.
Western Blotting Analysis: A representative lot detected PECAM-1 (CD31) in Western Blotting applications (Male, D., et. al. (1995). Immunology. 84(3):453-60).
ELISA Analysis: A representative lot detected PECAM-1 (CD31) in ELISA applications (Male, D., et. al. (1995). Immunology. 84(3):453-60; Williams, K.C., et. al. (1996). J Neurosci Res. 45(6):747-57).
Affects Function: A representative lot of PECAM-1 (CD31) Affected Function (Williams, K.C., et. al. (1996). J Neurosci Res. 45(6):747-57).
Immunohistochemistry Analysis: A representative lot detected PECAM-1 (CD31) in Immunohistochemistry applications (Williams, K.C., et. al. (1996). J Neurosci Res. 45(6):747-57).
Cell Structure
Quality
Immunohistochemistry Analysis: A 1:250 dilution of this antibody detected PECAM-1 (CD31) in human tonsil tissue.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service