Skip to Content
Merck
All Photos(1)

Key Documents

GF307

Sigma-Aldrich

β-NGF Protein, Human Recombinant Animal Free

The Nerve Growth Factor is a well characterized neurotropic protein that plays a critical role in the development of sympathetic & some sensory neurons in the peripheral nervous system. Manufactured using all non-animal reagents.

Synonym(s):

Nerve Growth Factor, NGF, BNGF, Beta NGF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77

Quality Level

Assay

>98% (SDS-PAGE and HPLC)

impurities

<0.1 μg/mg protein endotoxin (<1EU/μg)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

Nerve Growth Factor (β-NGF) is a well characterized neurotropic protein that plays a critical role in the development of sympathetic and some sensory neurons in the peripheral nervous system. In addition, NGF can also act in the central nervous system as a trophic factor for basal forebrain cholinergic neurons. NGF has also been shown to have biological effects on nonneuronal tissues. β-NGF is a neurotrophic factor structurally related to BDNF, NT-3 and NT-4. β-NGF is a potent neurotrophic factor that signals through its receptor β-NGFR, and plays a crucial role in the development and preservation of the sensory and sympathetic nervous systems. β-NGF also acts as a growth and differentiation factor for B lymphocytes and enhances B-cell survival. The functional form of human β-NGF is a noncovalently disulfide-linked homodimer, of two 13.5 kDa polypeptide monomers (238 total amino acid residues). The three disulfide bonds are required for biological activity.
Product Source: E. coli

Specificity

Cross Reactivty
Chicken, Hamster, Mouse, Rat

Application

Research Category
Neuroscience
Research Sub Category
Growth Factors & Receptors

Quality

Bioactivity assay: Assay 1: Determined by the dose-dependent stimulation of the proliferation of human TF-1 cells. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 106 units/mg.
Assay 2: Determined by its ability to stimulate chick E9 DRG neurite outgrowth. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 106 units/mg.

Physical form

Product is filtered through a 0.2 micron filter before lyophilization.
Recombinant animal free human β-NGF is manufactured using all non-animal reagents

Storage and Stability

Store at -20°C for up to 6 months from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service