Skip to Content
Merck
All Photos(1)

Key Documents

CC052

Sigma-Aldrich

Human Collagen Type II

liquid, 1 mg/mL, suitable for cell culture, used for gel formation

Synonym(s):

Collagen Type II, Human Collagen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.75

product name

Human Collagen Type II,

biological source

human

Quality Level

Assay

90% (Human collagen type II)

form

liquid

manufacturer/tradename

Chemicon®

concentration

1 mg/mL

technique(s)

cell culture | mammalian: suitable

impurities

<0.5% Non-collagen proteins
<1% Human collagen type IV
<1% Human collagen type V
<10% Human collagen type I
<10% Human collagen type III

input

sample type pancreatic stem cell(s)
sample type: human embryonic stem cell(s)
sample type epithelial cells
sample type induced pluripotent stem cell(s)
sample type mesenchymal stem cell(s)
sample type hematopoietic stem cell(s)
sample type neural stem cell(s)

solubility

water: soluble at 20 °C

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... COL2A1(1280)

General description

Type II collagen, a fibrillar collagen, is the main component of hyaline cartilage and constitutes 95% of the collagens and approximately 60% of cartilage’s dry weight. It is a homotrimer protein encoded by the gene COL2A1, mapped to human chromosome 12q13. Type two Collagen is also found in the nucleous pulposo of the intervertebral disc and the retina, sclera, and lens of the eye. Type IIA, is localized mainly to the vitreous body of the eye. Type IIB, is preferentially produced in adult cartilage tissue.  Collagen type II was purified by using differential salt precipitation, alcohol precipitation and DEAE chromatography. Multiple purification steps provide final preparations of collagen types in the native triple-helical form, free of any non-collagenous protein. Contamination of other collagen types in a standard preparation is lower than five percent.

Application

Human Collagen Type II has been used:
  • as an extracellular matrix (ECM) protein in microarray analysis, to coat on nitrocellulose film slides for probing and detection experiments
  • to study the role of Col2a1a in the neural crest during early eye development in zebrafish model of Stickler syndrome
  • as a calibration standard in the quantification of collagen II levels in scaffolds using an enzyme-linked immunosorbent assay (ELISA)-based assay

Biochem/physiol Actions

Type II collagen is commonly co-assembled with collagen XI, forming a complex that is covalently crosslinked to collagen IX and interacts with small leucine-rich proteoglycans. Collagen 2 imparts stability and strength to the tissue and maintains integrity and resilience under stress. Cleavage of Type II collagen occurs mainly through the action of matrix metalloproteinase collagenases, leading to the generation of well-known biomarkers such as the C-terminal telopeptide of Type II collagen and C2C. Mutations in type II collagen cause chondrodysplasia, leading to premature osteoarthritis.

Physical form

Purified protein. Liquid in 0.5M acetic acid. No preservative.

Storage and Stability

Aliquot and freeze. Maintain at -20°C up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular biology of cartilage matrix
LINE, et al.
AANA Journal (1993)
Type II collagen
Gudmann, et al.
Biochemistry of Collagens, Laminins and Elastin (2016)
Molecular biology of cartilage matrix
LINE, et al.
Journal of Separation Science (1993)
Zebrafish Model of Stickler Syndrome Suggests a Role for Col2a1a in the Neural Crest during Early Eye Development
Williams AL, et al,
Journal of developmental biology, 10(4) (2022)
Type II collagen
Gudmann, et al.
Biochemistry of Collagens, Laminins and Elastin (2016)

Articles

Extracellular matrix proteins such as laminin, collagen, and fibronectin can be used as cell attachment substrates in cell culture.

Extracellular matrix proteins such as laminin, collagen, and fibronectin can be used as cell attachment substrates in cell culture.

Extracellular matrix proteins such as laminin, collagen, and fibronectin can be used as cell attachment substrates in cell culture.

Extracellular matrix proteins such as laminin, collagen, and fibronectin can be used as cell attachment substrates in cell culture.

Protocols

This page covers the ECM coating protocols developed for four types of ECMs on Millicell®-CM inserts, Collagen Type 1, Fibronectin, Laminin, and Matrigel.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service