Skip to Content
Merck
All Photos(2)

Documents

CBL242

Sigma-Aldrich

Anti-Glucose Transporter GLUT-1 Antibody, CT

Chemicon®, from rabbit

Synonym(s):

Anti-Anti-CSE, Anti-Anti-DYT17, Anti-Anti-DYT18, Anti-Anti-DYT9, Anti-Anti-EIG12, Anti-Anti-GLUT, Anti-Anti-GLUT-1, Anti-Anti-GLUT1, Anti-Anti-GLUT1DS, Anti-Anti-HTLVR, Anti-Anti-PED, Anti-Anti-SDCHCN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat, human

manufacturer/tradename

Chemicon®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SLC2A1(6513)

Specificity

Specific for human glucose transporter 1 from muscle and adipose tissue. Does not cross-react with GLUT-4. The molecular weight of the glucose transporter precipitated by the serum is approximately 46 kDa.

Immunogen

12 amino acid peptide of the C-terminus of hGLUT-1 {LFHPLGADSQV} conjugated to KLH
Epitope: C-terminus

Application

Anti-Glucose Transporter GLUT-1 Antibody, C-terminus detects level of Glucose Transporter GLUT-1 & has been published & validated for use in IP & WB.
Research Category
Signaling
Research Sub Category
Insulin/Energy Signaling
Western blot analysis of GLUT-1 activity in adipose and muscle tissue (5μg/ml)

Immunoprecipitation of GLUT-1 (5μg/ml)

Optimal working dilutions must be determined by the end user.

Target description

46 kDa

Physical form

ImmunoAffinity Purified
The polyclonal is presented in phosphate buffered saline containing 10mM sodium azide. We recommend that each laboratory determine an optimum working titre for use in its particular application.

Storage and Stability

For use within 1 month of purchase store at +4°C, for long term storage aliquot antibody into small volumes and store at -20°C for up to one year.

Analysis Note

Control
Colorectal carcinoma

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angeliki Maria Nikolakopoulou et al.
The Journal of experimental medicine, 218(4) (2021-02-04)
The low-density lipoprotein receptor-related protein 1 (LRP1) is an endocytic and cell signaling transmembrane protein. Endothelial LRP1 clears proteinaceous toxins at the blood-brain barrier (BBB), regulates angiogenesis, and is increasingly reduced in Alzheimer's disease associated with BBB breakdown and neurodegeneration.
Fei Liu et al.
The European journal of neuroscience, 37(10), 1714-1725 (2013-02-26)
Deposition of β -amyloid (Aβ) peptides, cleavage products of β-amyloid precursor protein (APP) by β-secretase-1 (BACE1) and γ-secretase, is a neuropathological hallmark of Alzheimer's disease (AD). γ-Secretase inhibition is a therapeutical anti-Aβ approach, although changes in the enzyme's activity in
M G Alves et al.
British journal of pharmacology, 171(4), 1033-1042 (2013-11-23)
Metformin is commonly used to treat type 2 diabetes (T2D). While new clinical applications have been ascribed to metformin, including treatment of anovulatory infertility, its effects on male reproduction have not been investigated. The Sertoli cell (SC) is crucial for
Jiaqi Ren et al.
The Journal of physiology, 599(20), 4705-4724 (2021-09-07)
Restriction of fetal substrate supply has an adverse effect on surfactant maturation in the lung and thus affects the transition from in utero placental oxygenation to pulmonary ventilation ex utero. The effects on surfactant maturation are mediated by alteration in
Giuseppe Barisano et al.
The Journal of experimental medicine, 219(11) (2022-08-31)
Apolipoprotein E4 (APOE4), the main susceptibility gene for Alzheimer's disease, leads to blood-brain barrier (BBB) breakdown in humans and mice. Remarkably, BBB dysfunction predicts cognitive decline and precedes synaptic deficits in APOE4 human carriers. How APOE4 affects BBB and synaptic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service