Skip to Content
Merck
All Photos(1)

Key Documents

CBL1512

Sigma-Aldrich

Anti-Integrin αM [CD11b] Antibody, clone OX-42

clone OX-42, Chemicon®, from mouse

Synonym(s):

CD11b

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

OX-42, monoclonal

species reactivity

rat

packaging

antibody small pack of 25 μg

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

Related Categories

General description

Integrin alpha M/beta 2 is implicated in various adhesive interactions of monocytes, macrophages and granulocytes as well as in mediating the uptake of complement-coated particles. It is identical with CR 3, the receptor for the iC3b fragment of the third complement component. It probably recognizes the R-G-D peptide in C3b. Integrin alpha M/beta 2 is also a receptor for fibrinogen, factor X and ICAM1. It recognizes P1 and P2 peptides of fibrinogen gamma chain.

Specificity

Recognizes most macrophages (including resident peritoneal and activated macrophages), kupffer cells, but only about 35% of alveolar macrophages. The antibody also labels dendritic cells extensively, granulocytes and cells with the morphology of microglia in brain. The antibody precipitates three polypeptides of 150, 103 and 95 KDa. The antibody inhibits complement mediated rosettes and is probably the rat equivalent of the human receptor for IC3b called CR3.
FUSION PARTNER: NSO myeloma cell line.

Immunogen

Rat peritoneal macrophages

Application

Detect Integrin αM [CD11b] using this Anti-Integrin αM CD11b Antibody, clone OX-42 validated for use in FC, IH, IH(P) & IP.
Research Category
Cell Structure
Research Sub Category
Integrins

Physical form

Format: Purified
Protein A purified
The monoclonal is presented in phosphate buffered saline containing 10mM sodium azide.

Storage and Stability

For use within 1 month of purchase store at +4°C, for long term storage aliquot antibody into small volumes and store at -20°C.

Analysis Note

Control
Brain tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Wang et al.
Molecular medicine reports, 7(4), 1137-1142 (2013-02-22)
Postoperative cognitive dysfunction (POCD), common in elderly patients, refers to a decline in cognitive function following surgery, which may persist or even evolve into Alzheimer's disease (AD). Despite great efforts, the mechanism of POCD remains unclear. In the present study
Reduction of ?-amyloid deposits by ?-secretase inhibitor is associated with the attenuation of secondary damage in the ipsilateral thalamus and sensory functional improvement after focal cortical infarction in hypertensive rats.
Zhang, Y; Xing, S; Zhang, J; Li, J; Li, C; Pei, Z; Zeng, J
Journal of Cerebral Blood Flow and Metabolism null
Elisa Torres et al.
Journal of neuroinflammation, 8, 53-53 (2011-05-21)
3,4-Methylenedioxymethamphetamine (MDMA) produces a neuroinflammatory reaction in rat brain characterized by an increase in interleukin-1 beta (IL-1β) and microglial activation. The CB2 receptor agonist JWH-015 reduces both these changes and partially protects against MDMA-induced neurotoxicity. We have examined MDMA-induced changes
HDAC inhibitor increases histone H3 acetylation and reduces microglia inflammatory response following traumatic brain injury in rats.
Zhang, B; West, EJ; Van, KC; Gurkoff, GG; Zhou, J; Zhang, XM; Kozikowski, AP; Lyeth, BG
Brain Research null
Jee Youn Lee et al.
Journal of ginseng research, 43(1), 58-67 (2019-01-22)
Diabetic neuropathy is one of the most devastating ailments of the peripheral nervous system. Neuropathic pain develops in ∼30% of diabetics. Here, we examined the suppressive effect of GS-KG9 on neuropathic pain induced by streptozotocin (STZ). Hyperglycemia was induced by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service