Skip to Content
Merck
All Photos(1)

Key Documents

AGX840

Sigma-Aldrich

Recombinant SARS-CoV-2 Nucleocapsid Protein (whole length)

suitable for ELISA

Synonym(s):

2019-nCoV, SARS-CoV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51201600
NACRES:
NA.26

recombinant

expressed in E. coli

Quality Level

form

liquid

storage condition

avoid repeated freeze/thaw cycles

technique(s)

ELISA: suitable

shipped in

dry ice

General description

This e. coli recombinant SARS-CoV-2 Nucleocapsid Protein (whole length), Cat. No. AGX840 is tested for use in ELISA.

Specificity

SARS CoV2

Target description

Coronavirus disease 2019 (COVID-19) is an infectious disease caused by the virus strain severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), a virus closely related to the SARS virus. A SARS-CoV-2 nucleocapsid antigen can be used to support the detection of SARS-CoV-2 (COVID-19, 2019-nCoV).

Physical form

Purified recombinant protein in 0.01M PBS, pH7.4

Reconstitution

Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service