Skip to Content
Merck
All Photos(6)

Key Documents

AB2253

Sigma-Aldrich

Anti-Doublecortin Antibody

serum, from guinea pig

Synonym(s):

Anti-Doublecortin Antibody, AB2253 | Sigma-Aldrich

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

guinea pig

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

immunofluorescence: suitable (Frozen tissue sections)
immunohistochemistry: suitable (Paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DCDC1(341019)

General description

Anti-Doublecortin, Cat. No. AB2253, is a guinea pig polyclonal antibody that detects Doublecortin and is tested for use in Immunohistochemistry (Paraffin) and Immunofluorescence, and Western Blotting.
Target molecular weight ~42 kDa observed;40.61 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Specificity

Reactivity with other species has not been determined.
This Guinea pig polyclonal antibody detects doublecortin in murine and rat species. It targets an epitope with in 17 amino acids from the C-terminal region.

Immunogen

Epitope: C-Terminus
Recombinant Peptide.

Application

Anti-Doublecortin Antibody has been used in immunohistochemistry and immunofluorescence staining.

Biochem/physiol Actions

Doublecortin (DCX) is identified as an important marker for neurogenesis. It serves as a valuable tool for studying and comparing the process of neurogenesis across different species.

Quality

Evaluated by Western Blotting in rat brain tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Doublecortin in rat brain tissue lysate.

Target description

~45 kDa observed; 40.57 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: AB5910

Physical form

GST Depletion
GST-Depleted Guinea Pig Sera with 0.05% Sodium Azide

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Rat Brain lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacques Balthazart et al.
Brain, behavior and evolution, 84(1), 1-4 (2014-07-19)
Commentary on “Evaluating the Predictive Value of Doublecortin as a Marker for Adult Neurogenesis in Canaries (Serinus canaria)” by Michiel Vellema, Moritz Hertel, Susan L. Urbanus, Annemie Van der Linden, and Manfred Gahr (Journal of Comparative Neurology 2014, 522 :
Embryonic Nkx2. 1-expressing neural precursor cells contribute to the regional heterogeneity of adult V-SVZ neural stem cells
Delgado RN and Lim DA
Developmental Biology, 407(2), 265-274 (2015)
Tania Campos-Ordoñez et al.
Experimental neurology, 261, 236-244 (2014-05-27)
Hydrocephalus can develop secondarily to a disturbance in production, flow and/or absorption of cerebrospinal fluid. Experimental models of hydrocephalus, especially subacute and chronic hydrocephalus, are few and limited, and the effects of hydrocephalus on the subventricular zone are unclear. The
Combination treatment with ethyl pyruvate and IGF-I exerts neuroprotective effects against brain injury in a rat model of neonatal hypoxic-ischemic encephalopathy
Rong Z, et al.
International Journal of Molecular Medicine, 36(1), 195-203 (2015)
Hanadie Yousef et al.
Oncotarget, 6(14), 11959-11978 (2015-05-25)
Stem cell function declines with age largely due to the biochemical imbalances in their tissue niches, and this work demonstrates that aging imposes an elevation in transforming growth factor β (TGF-β) signaling in the neurogenic niche of the hippocampus, analogous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service