Skip to Content
Merck
All Photos(1)

Key Documents

8.52112

Sigma-Aldrich

Fmoc-Lys(Me₃Cl)-OH

Novabiochem®

Synonym(s):

Fmoc-Lys(Me₃Cl)-OH, N-α-Fmoc-N-ε-trimethyl-N-L-lysine · chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H31ClN2O4
CAS Number:
Molecular Weight:
446.97
UNSPSC Code:
12352209
NACRES:
NA.22

Quality Level

product line

Novabiochem®

Assay

≥95% (TLC)
≥95.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

InChI

1S/C24H30N2O4.ClH/c1-26(2,3)15-9-8-14-22(23(27)28)25-24(29)30-16-21-19-12-6-4-10-17(19)18-11-5-7-13-20(18)21;/h4-7,10-13,21-22H,8-9,14-16H2,1-3H3,(H-,25,27,28,29);1H/t22-;/m0./s1

InChI key

XUJRNPVABVHOAJ-FTBISJDPSA-N

General description

A novel derivative for the introduction of trimethyl-lysine during Fmoc SPPS. Ref [1] contains methods and protocols for the synthesis of arrays of histone-related peptides containing methylated arginine and lysine-residues.


Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Overcoming Aggregation in Fmoc SPPS

Literature references

[1] S. Rothbart, et al. (2012) Methods Enzymol., 512, 107.

Linkage

Replaces: 04-12-1270

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Purity (TLC(CMA1)): ≥ 95 %
Assay (HPLC, area%): ≥ 95.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
LC/MS: passes test

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Unnatural amino acids, the non-proteinogenic amino acids that either occur naturally or are chemically synthesized, are becoming more and more important as tools for modern drug discovery research.

Unnatural amino acids, the non-proteinogenic amino acids that either occur naturally or are chemically synthesized, are becoming more and more important as tools for modern drug discovery research.

Unnatural amino acids, the non-proteinogenic amino acids that either occur naturally or are chemically synthesized, are becoming more and more important as tools for modern drug discovery research.

Unnatural amino acids, the non-proteinogenic amino acids that either occur naturally or are chemically synthesized, are becoming more and more important as tools for modern drug discovery research.

Protocols

We provide an overview of our available reagents, together with recommendations and details of their use for synthesis of peptides containing post-translationally modified amino acids.

We provide an overview of our available reagents, together with recommendations and details of their use for synthesis of peptides containing post-translationally modified amino acids.

We provide an overview of our available reagents, together with recommendations and details of their use for synthesis of peptides containing post-translationally modified amino acids.

We provide an overview of our available reagents, together with recommendations and details of their use for synthesis of peptides containing post-translationally modified amino acids.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service