Skip to Content
Merck
All Photos(1)

Key Documents

8.20095

Sigma-Aldrich

5-Amino-2-hydroxybenzoic acid

for synthesis

Synonym(s):

5-Amino-2-hydroxybenzoic acid, 5-Aminosalicylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
5-(NH2)-2-(OH)C6H3COOH
CAS Number:
Molecular Weight:
153.14
MDL number:
UNSPSC Code:
12352106
EC Index Number:
201-919-1

form

powder

autoignition temp.

280 °C

potency

2800 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rat)

pH

4.0-4.1 (20 °C, 0.8 g/L in H2O)

mp

280 °C (decomposition)

solubility

0.84 g/L

bulk density

300 kg/m3

storage temp.

2-30°C

InChI

1S/C7H7NO3/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3,9H,8H2,(H,10,11)

InChI key

KBOPZPXVLCULAV-UHFFFAOYSA-N

Analysis Note

Assay (HClO₄): ≥ 97.0 %
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(C)

280 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service