Skip to Content
Merck
All Photos(2)

Key Documents

8.14442

Sigma-Aldrich

3,3,5-Trimethylcyclohexanol

(mixture of cis- and trans- isomers) for synthesis

Synonym(s):

3,3,5-Trimethylcyclohexanol, Homomenthol, Dihydroisophorol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18O
CAS Number:
Molecular Weight:
142.24
MDL number:
UNSPSC Code:
12352104
EC Index Number:
204-122-7

vapor pressure

1 hPa ( 50 °C)

form

crystals

autoignition temp.

375 °C (DIN 51794)

potency

3250 mg/kg LD50, oral (Rat)
2430 mg/kg LD50, skin (Rabbit)

expl. lim.

1.5 % (v/v)

pH

7 (20 °C, 1.8 g/L in H2O)

bp

193-196 °C/1013 hPa

mp

30-32 °C

transition temp

flash point 81 °C (DIN 51758)

solubility

1.8 g/L

density

0.86 g/cm3 at 60 °C

storage temp.

2-30°C

SMILES string

OC1CC(CC(C)(C1)C)C

InChI

1S/C9H18O/c1-7-4-8(10)6-9(2,3)5-7/h7-8,10H,4-6H2,1-3H3

InChI key

BRRVXFOKWJKTGG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay isomers (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 28 °C
Melting range (upper value): ≤ 33 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(C)

88 °C - open cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service