Skip to Content
Merck
All Photos(1)

Key Documents

8.07993

Sigma-Aldrich

Sulfolane

for synthesis

Synonym(s):

Sulfolane, Tetrahydrothiophene 1,1-dioxide, Tetramethylene sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2S
CAS Number:
Molecular Weight:
120.17
MDL number:
UNSPSC Code:
12352119
EC Index Number:
204-783-1
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.035 hPa ( 30 °C)

Quality Level

Assay

≥99.0% (GC)

form

solid

potency

2006 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

bp

285 °C/1013 hPa

mp

26 °C

transition temp

flash point 177 °C

solubility

soluble ≥100 g/L

density

1.26 g/cm3 at 30 °C

storage temp.

2-30°C

SMILES string

[S]1(=O)(=O)CCCC1

InChI

1S/C4H8O2S/c5-7(6)3-1-2-4-7/h1-4H2

InChI key

HXJUTPCZVOIRIF-UHFFFAOYSA-N

Application


  • Comparative Kinetics Study of CO2 Absorption into Aqueous MDEA/PZ, MDEA/Sulfolane, and PZ/Sulfolane Blended Systems Solution, Using Stopped-Flow Technique.: This research investigates the kinetics of CO2 absorption in various blended aqueous solutions, including those with sulfolane, using the stopped-flow technique. The study provides insights into the efficiency of CO2 capture using sulfolane-based systems (Abdelmouiz et al., 2022).

  • Experimental Measurement of Carbon Dioxide Solubility in Aqueous N-Methyldiethanolamine + 2-(2-Aminoethylamino) Ethanol + Sulfolane and Diethanolamine + 2-(2-Aminoethylamino) Ethanol + Sulfolane Hybrid Solvents at Various Temperatures and High Pressure.: This paper presents experimental data on the solubility of CO2 in hybrid solvents containing sulfolane, demonstrating its effectiveness in CO2 capture under different conditions (Asadi et al., 2020).

  • Photocatalytic Degradation of Sulfolane Using a LED-Based Photocatalytic Treatment System.: This study explores the degradation of sulfolane in aqueous media using a photocatalytic system with titanium dioxide and reduced graphene oxide, highlighting the potential of LED-based photocatalysis for environmental cleanup (Dharwadkar et al., 2021).

  • Temperature-Related Corrosion Resistance of AISI 1010 Carbon Steel in Sulfolane.: This research evaluates the corrosion resistance of carbon steel in sulfolane at various temperatures, providing valuable data on the material compatibility in industrial applications where sulfolane is used as a solvent (Kubisztal et al., 2020).

  • Water-Induced Corrosion Damage of Carbon Steel in Sulfolane.: The paper investigates the effects of water contamination in sulfolane on the corrosion of carbon steel, emphasizing the importance of controlling water levels to prevent industrial equipment degradation (Kubisztal et al., 2020).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Water (K. F.): ≤ 0.20 %
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

350.6 °F - closed cup

Flash Point(C)

177 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service