Skip to Content
Merck
All Photos(1)

Key Documents

8.00762

Sigma-Aldrich

Terephthalic acid

for synthesis

Synonym(s):

Terephthalic acid, 1,4-Benzenedicarboxylic acid, p-Phthalic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,4-(COOH)2
CAS Number:
Molecular Weight:
166.13
MDL number:
UNSPSC Code:
12352106
EC Index Number:
202-830-0
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (acidimetric)

form

crystalline

autoignition temp.

496 °C

potency

>6400 mg/kg LD50, oral (Rat)

mp

402 °C (sublimed)

solubility

15 mg/L (experimental)

density

1.51 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[O-]C(=O)c1ccc(cc1)C(=O)[O-].[H+].[H+]

InChI

1S/C8H6O4/c9-7(10)5-1-2-6(4-3-5)8(11)12/h1-4H,(H,9,10)(H,11,12)

InChI key

KKEYFWRCBNTPAC-UHFFFAOYSA-N

Application


  • Biotransformation of p-xylene into terephthalic acid by engineered Escherichia coli: This study explores an alternative production method for terephthalic acid (TPA) that could be less energy-intensive and safer than traditional methods. It reports the successful bioconversion of p-xylene into TPA using genetically engineered E. coli, highlighting a potential sustainable approach to TPA production (Luo & Lee, 2017).

  • Synthesis and verification of biobased terephthalic acid from furfural: This research presents a novel synthetic pathway for producing TPA from biomass-derived furfural. It represents a significant step towards sustainable chemical production, providing a greener alternative to petrochemical routes (Tachibana, Kimura & Kasuya, 2015).

  • Studies on the solubility of terephthalic acid in ionic liquids: Focused on identifying better solvents for TPA, this study investigates the solubility of TPA in various ionic liquids. The findings could lead to improved processing techniques for TPA in industrial applications (Matuszek et al., 2019).

  • Engineering terephthalic acid product from recycling of PET bottles: This study explores the depolymerization of PET into TPA and ethylene glycol, highlighting a sustainable approach to recycle PET waste and reduce reliance on petrochemicals (Lee, Chiu & Lee, 2021).

  • Biobased terephthalic acid technologies - a literature review: Reviews various biobased approaches to produce TPA, focusing on the potential of renewable feedstocks to replace traditional petrochemical routes. This comprehensive review discusses different biotechnological and chemical methods to synthesize TPA from bio-based sources (Collias et al., 2014).

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service