Skip to Content
Merck
All Photos(1)

Documents

71285-M

Millipore

BCA Protein Assay Kit

The BCA protein assay is a simple and reliable protein quantification method.

Synonym(s):

BCA Protein Quantification, Bicinchoninic Acid Assay, Protein Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.84

manufacturer/tradename

Novagen®

storage condition

OK to freeze

storage temp.

10-30°C

General description

The BCA protein assay is a simple and reliable protein quantification method.
The BCA protein assay is based on a biuretreaction, which is the reduction of Cu2+ toCu+ by proteins in an alkaline solution withconcentration-dependent detection of themonovalent copper ions. Bicinchoninic acidis a chromogenic reagent that chelates thereduced copper, producing a purple complexwith strong absorbance at 562 nm (Smith1985, Wiechelman 1988). This assay can beused to quantify protein concentration with awide variety of samples and can be performedin minutes.

The Novagen® BCA Protein Assay Kit canbe used to determine protein concentrationin the range of 20-2000 µg/ml in either astandard assay or microassay configuration.Kit components are sufficient to complete500 standard-size reactions (50 µl proteinsample plus 1 ml reagent) or 2500 micro-scalereactions (25 µl protein sample plus 200 µlreagent). A BSA standard (2 mg/ml) is providedfor convenient and reliable preparation ofstandard curves.

This assay is robust and can be performedin the presence of many compounds. Somereagents, including chelating agents, strongacids or bases, and reducing agents, interferewith the reduction and chelating reactions onwhich this assay depends (Brown 1989). TheBCA assay is compatible with the followingNovagen protein extraction and lysis reagents:BugBuster Protein Extraction Reagent,PopCulture Reagent, CytoBuster ProteinExtraction Reagent, Reportasol ExtractionBuffer, and Insect PopCulture Reagent. Optionsfor the removal or dilution of interferingsubstances are described in the kit literature.

Components

•500 mlBCA Solution

•15 ml4% Cupric Sulfate

•3 x 1 mlBSA Standard, 2 mg/ml

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Other Notes

Smith, P.K., et al. 1985. Anal. Biochem.150, 76.

Kessler, R. J. and Fanestil, D. D. 1986. Anal. Biochem 159, 138.

Wiechelman, K., et al. 1988. Anal. Biochem.175, 231.

Brown, R., et al. 1989. Anal. Biochem.180, 136.

Legal Information

Use of the BCA protein assay is permitted for research purposes only.
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Shuhan Liu et al.
Oxidative medicine and cellular longevity, 2021, 7674565-7674565 (2021-12-18)
Cysteine metabolism plays a critical role in cancer cell survival. Cysteine depletion was reported to inhibit tumor growth and induce pancreatic cancer cell ferroptosis. Nevertheless, the effect of cysteine depletion in chronic myeloid leukemia (CML) remains to be explored. In
Roberta Gonnella et al.
Microbes and infection, 22(10), 585-591 (2020-09-04)
Viral egress and autophagy are two mechanisms that seem to be strictly connected in Herpesviruses's biology. Several data suggest that the autophagic machinery facilitates the egress of viral capsids and thus the production of new infectious particles. In the Herpesvirus
Fabien Wauquier et al.
Nutrients, 13(8) (2021-08-28)
The aging of our population is accompanied by an increased prevalence of chronic diseases. Among those, liver, joint and adipose tissue-related pathologies have a major socio-economic impact. They share common origins as they result from a dysregulation of the inflammatory
Abderrahim Benmoussa et al.
Bio-protocol, 10(11), e3636-e3636 (2021-03-05)
Milk is a complex fluid that contains various types of proteins and extracellular vesicles (EVs). Some proteins can mingle with EVs, and interfere with their isolation. Among these proteins, caseins form micelles of a size comparable to milk EVs, and
Abdullah S Shatoor et al.
Saudi journal of biological sciences, 28(12), 7012-7021 (2021-12-07)
This study evaluated the protective effect of astaxanthin (ASX) against high-fat diet (HFD)-induced cardiac damage and fibrosis in rats and examined if the mechanism of protection involves modulating SIRT1. Rat were divided into 5 groups (n = 10/group) as: 1) control: fed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service