Skip to Content
Merck
All Photos(1)

Key Documents

616374

Sigma-Aldrich

TRAIL, Human, Recombinant, E. coli

Synonym(s):

Apo2L, Human, Recombinant, E. coli, TNF-Related Apoptosis-Inducing Ligand, Human, Recombinant, E. coli

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

Assay

≥95% (SDS-PAGE)

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

impurities

<0.1 EU/μg endotoxin (EU/μg TRAIL)

shipped in

wet ice

storage temp.

−70°C

General description

Recombinant, human TRAIL (amino acids 114-281) expressed in E. coli. Ligand for TRAIL receptors DR4 and DR5. Displays potent antitumor activity against selected targets and induces apoptosis in a number of transformed cell lines, but not in normal tissues.

Biochem/physiol Actions

Induces apoptosis in U937 cells at 1 μg/ml within 5 h.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 500 mM NaCl, 10 mM Na₂HPO₄, 2.7 mM KCl, 2 mM KH₂PO₄, 0.1 mM DTT, ≤10% glycerol.

Reconstitution

Following initial thaw, aliquot and freeze

(-70°C).

Other Notes

Lin, Y., et al. 2000. Mol. Cell. Biol.20, 6638.
Mitsaides, N., et al. 2000. Cancer Res.60, 4122.
Griffith, T.S., et al. 1999. J. Exp. Med.189, 1343.
Mariani, S.M., et al. 1997. J. Cell Biol.137, 221.
Pan, G., et al. 1997. Science276, 111.
Pan, G., et al. 1997. Science277, 815.
Sheridan, J.P., et al. 1997. Science277, 818.
Pitti, R.M., et al. 1996. J. Biol. Chem.271, 12687.
Wiley, S.R., et al. 1995. Immunity3, 673.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oliver H Voss et al.
Cell death & disease, 12(4), 287-287 (2021-03-19)
Tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) selective killing of cancer cells underlines its anticancer potential. However, poor tolerability and resistance underscores the need to identify cancer-selective TRAIL-sensitizing agents. Apigenin, a dietary flavonoid, sensitizes lung cancer cell lines to TRAIL. It

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service