Skip to Content
Merck
All Photos(1)

Key Documents

5.38337

Sigma-Aldrich

PDE9 Inhibitor, PF-04447943

Synonym(s):

PDE9 Inhibitor, PF-04447943, 6-((3S,4S)-4-methyl-1-(pyrimidin-2-ylmethyl)pyrrolidin-3-yl)-1-(tetrahydro-2H-pyran-4-yl)-1H-pyrazolo[3,4-d]pyrimidin-4(5H)-one, PF04447943, PF 04447943, Phosphodiesterase 9A Inhibitor, PF-04447943

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H25N7O2
CAS Number:
Molecular Weight:
395.46
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

storage temp.

−20°C

InChI

1S/C20H25N7O2/c1-13-10-26(12-17-21-5-2-6-22-17)11-16(13)18-24-19-15(20(28)25-18)9-23-27(19)14-3-7-29-8-4-14/h2,5-6,9,13-14,16H,3-4,7-8,10-12H2,1H3,(H,24,25,28)/t13-,16-/m1/s1

InChI key

IWXUVYOOUMLUTQ-CZUORRHYSA-N

General description

A cell-permeable, blood-brain barrier permeant, pyrazolo[3,4 d]pyrimidinone compound that acts as a highly potent, selective, and active site-targeting inhibitor of phosphodiesterase 9A (PDE9A; Ki = 2.8 nM, 4.5 nM, and 18 nM for human, rhesus monkey, and rat recombinant PDE9A, respectively). Exhibits high selectivity over other phosphodiesterases (Ki = 8.6, 99, 50, 29, 14.9, 5.3, 75, 50, 51.2, and 80 µM for PDE1, PDE2A3, PDE3A, PDE4A, PDE5A, PDE6C, PDE7A2, PDE8A, PDE10, PDE11, respectively). Does not affect the activity of 60 other enzymes and receptors. Shown to diminish ANP-stimulated cGMP levels in human embryonic kidney cells expressing rhesus PDE9A2 (IC50 = 375 nM). Shown to improve performance in novel object recognition in rats (~ 3 mg/kg, p.o.). Exhibits desirable pharmacokinetic properties with t1/2 = 4.9 h, Tmax = 0.3 h in rat.
A cell-permeable, blood-brain barrier permeant, pyrazolo[3,4 d]pyrimidinone compound that acts as a highly potent, selective, and active site-targeting inhibitor of phosphodiesterase 9A (PDE9A; Ki = 2.8 nM, 4.5 nM, and 18 nM for human, rhesus monkey, and rat recombinant PDE9A, respectively). Exhibits high selectivity over other phosphodiesterases (Ki = 8.6, 99, 50, 29, 14.9, 5.3, 75, 50, 51.2, and 80 µM for PDE1, PDE2A3, PDE3A, PDE4A, PDE5A, PDE6C, PDE7A2, PDE8A, PDE10, PDE11, respectively). Does not affect the activity of 60 other enzymes and receptors. Shown to diminish ANP-stimulated cGMP levels in human embryonic kidney cells expressing rhesus PDE9A2 (IC50 = 375 nM). Shown to improve performance in novel object recognition in rats (~ 3 mg/kg, p.o.). Exhibits desirable pharmacokinetic properties with t1/2 = 4.9 h, Tmax = 0.3 h in rat.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PDE9
Reversible: yes
Target IC50: 2.8 nM, 4.5 nM and 18 nM for human, rhesus and rat PDE9A inhibition respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Lee, D., et al. 2015. Nature.519, 472

Verhoest, P. R., et al. 2012. J. Med. Chem.55, 9045.

Hutson, P.H., et al. 2011. Neuropharmacology.61, 665.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service