Skip to Content
Merck
All Photos(2)

Key Documents

356352-M

Millipore

Glycerol

Molecular Biology Grade

Synonym(s):

Glycerol, 1,2,3-Propanetriol, Glycerin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
92.09
Beilstein:
635685
MDL number:
UNSPSC Code:
50161901
NACRES:
NA.79
grade:
Molecular Biology
Assay:
≥99% (titration)
technique(s):
electrophoresis: suitable
bp:
182 °C/20 mmHg (lit.)
vapor pressure:
<1 mmHg ( 20 °C)

grade

Molecular Biology

Quality Level

vapor density

3.1 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

description

Merck USA index - 14, 4485

Assay

≥99% (titration)

form

liquid

autoignition temp.

698 °F

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

technique(s)

electrophoresis: suitable

refractive index

n20/D 1.474 (lit.)

bp

182 °C/20 mmHg (lit.)

mp

20 °C (lit.)

density

1.25 g/mL (lit.)

cation traces

heavy metals: ≤0.0005%

absorption

≤0.02 at 280 nm in H2O at 0.5 M
≤0.07 at 260 nm in H2O at 0.5 M

foreign activity

DNases, proteases, RNases, none detected

storage temp.

10-30°C

SMILES string

OCC(O)CO

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

PEDCQBHIVMGVHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Obtained from oils and fats as a biproduct in the manufacturing of soaps and fatty acids (absorbance: ≤0.07 at 260 nm; ≤0.02 at 280 nm).

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

390.2 °F - Pensky-Martens closed cup

Flash Point(C)

199 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service