Skip to Content
Merck
All Photos(1)

Key Documents

324412

Sigma-Aldrich

Dynamin Inhibitor III, Phthaladyn-23

Synonym(s):

Dynamin Inhibitor III, Phthaladyn-23, 4-Chloro-2-((2-(3-nitrophenyl)-1,3-dioxo-2,3-dihydro-1H-isoindole-5-carbonyl)-amino)-benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H12ClN3O7
Molecular Weight:
465.80
UNSPSC Code:
12352200
NACRES:
NA.28

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light yellow to beige

solubility

DMSO: 7 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable phthalimide compound that is reported to inhibit dynamin GTPase activity (IC50 = 17.4 and 63 µM using purified sheep brain dynamin I and recombinant dynamin II, respectively) in a GTP-competitive manner. Shown to block K+-induced SVE (synaptic vesicle endocytosis) of FM4-64 dye into rat brain synaptosomes (IC50 = 12.9 µM), while being ineffective at 30 µM against RME (receptor-mediated endocytosis) of transferrin into human osteosarcoma epithelial U2OS cells, presumably due to poor dynamin II inhibitory potency and/or rapid efflux and/or limited permeability in this cell line.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Odell, L.R., et al. 2010. J. Med. Chem.53, 5267.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service