Skip to Content
Merck
All Photos(1)

Documents

253300

Sigma-Aldrich

Deltamethrin

Synthetic type II pyrethroid. Potent inhibitor of calcineurin. Causes an increase in neurotransmitter release at synapses, and increases intrasynaptosomal Ca2+ levels.

Synonym(s):

Deltamethrin, (S)-α-Cyano-3-phenoxybenzyl(1R)- cis-3-(2,2-dibromovinyl)-2,2-dimethylcyclopropanecarboxylate, (S)-α-Cyano-3-phenoxybenzyl(1R)-cis-3-(2,2-dibromovinyl)-2,2-dimethylcyclopropanecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H19Br2NO3
CAS Number:
Molecular Weight:
505.20
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

ethanol: 1 mg/mL
DMSO: 15 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C22H19Br2NO3/c1-22(2)17(12-19(23)24)20(22)21(26)28-18(13-25)14-7-6-10-16(11-14)27-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3/t17-,18+,20-/m0/s1

InChI key

OWZREIFADZCYQD-NSHGMRRFSA-N

General description

Synthetic type II pyrethroid insecticide. Potent inhibitor of calcineurin (protein phosphatase 2B, IC50 = 100 pM). May also have varied effects on ion channels. Causes an increase in neurotransmitter release at synapses and also increases intrasynaptosomal Ca2+ levels.
Synthetic type II pyrethroid. Potent inhibitor of calcineurin (protein phosphatase 2B; IC50 = 100 pM). May also have varied effects on ion channels. Causes an increase in neurotransmitter release at synapses, and increases intrasynaptosomal Ca2+ levels.

Biochem/physiol Actions

Cell permeable: no
Primary Target
protein phosphatase 2B
Product does not compete with ATP.
Reversible: no
Target IC50: 100 pM against calcineurin (protein phosphatase 2B)

Warning

Toxicity: Highly Toxic (H)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C. Avoid acid or alkali conditions.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Richter, A., et al. 1995. Biochem. Pharmacol.49, 367.
Enan, E., and Matsumura, F. 1993. Biochem. Pharmacol. 45, 703.
Enan, E., and Matsumura, F. 1992. Biochem. Pharmacol.43, 1777.
Narahasi, T. 1992. Trends Pharmacol. Sci.13, 236.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Richter et al.
Biochemical pharmacology, 49(3), 367-373 (1995-01-31)
The ability of the immunosuppressive agent FK506 to affect growth of the epidermal growth factor-receptor (EGF-R) overexpressing cell line, A431, was compared with that of the structurally unrelated immunosuppressive compound, cyclosporin A (CyA). Both were shown to inhibit growth, although
T Narahashi
Trends in pharmacological sciences, 13(6), 236-241 (1992-06-01)
The mechanisms of action of neuroactive insecticides on the nervous system has been studied for many years. It is now well established that severe neurological symptoms of poisoning with pyrethroids and DDT in mammals and insects are the result of
E Enan et al.
Biochemical pharmacology, 45(3), 703-710 (1993-02-09)
We have investigated the effects of a type II pyrethroid insecticide, deltamethrin, on changes in the protein phosphorylation pattern associated with neurotransmitter release in rat brain synaptosomal preparations. Deltamethrin was found to stimulate directly the activity of the protein kinase
E Enan et al.
Biochemical pharmacology, 43(8), 1777-1784 (1992-04-15)
The inhibitory action of synthetic pyrethroids and some chlorinated hydrocarbon insecticides on the neural calcium-calmodulin-dependent protein phosphatase, calcineurin, was studied using one radiotracer and two colorimetric methods. It was found that all insecticidal Type II pyrethroids (cypermethrin, deltamethrin and fenvalerate)
Guey-Ying Liao et al.
SLAS discovery : advancing life sciences R & D, 28(3), 88-94 (2023-02-27)
Deficiencies in brain-derived neurotrophic factor (BDNF) have been linked to several brain disorders, making compounds that can boost neuronal BDNF synthesis attractive as potential therapeutics. However, a sensitive and quantitative BDNF assay for high-throughput screening (HTS) is still missing. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service