Skip to Content
Merck
All Photos(1)

Key Documents

219467

Sigma-Aldrich

Cathepsin V, His•Tag®, Human, Recombinant, NSO Cells

recombinant human, expressed in NSO cells

Synonym(s):

Cathepsin L2, Cathepsin U

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352202
NACRES:
NA.77

recombinant

expressed in NSO cells

Quality Level

Assay

≥95% (SDS-PAGE)

form

frozen liquid

specific activity

≥300 units/μg

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

impurities

≤1.0 EU/μg Endotoxins (EU/μg Cathepsin V)

shipped in

wet ice

storage temp.

−70°C

General description

Recombinant, human procathepsin V (amino acids 18-334) fused at the N-terminus to a CD33 signal sequence (amino acids 1-16) and at the C-terminus to a His•Tag sequence and expressed in NSO cells. Cat V is predominantly expressed in normal thymus, testis, and corneal epithelium. It is involved in the invariant chain degradation in thymic epithelial cells and its overexpression has been detected in myasthenia gravis. Abnormally wide spread expressions in mammary gland and colon have also been reported in breast and colonrectal carcinomas. Although it shows high amino acid identity with cathepsin L (Cat. No. 219382), these two cathepsins have very different tissue distribution and substrate specificity. As a result of glycosylation, this recombinant enzyme migrates ~40 kDa on SDS-PAGE under either reducing or non-reducing conditions. Requires activation prior to use.
Recombinant, human procathepsin V (amino acids 18-334) fused at the N-terminus to a CD33 signal sequence (amino acids 1-16) and at the C-terminus to a His•Tag sequence and expressed in NSO cells. Cat V is predominantly expressed in normal thymus, testis, and corneal epithelium. It is involved in the invariant chain degradation in thymic epithelial cells and its overexpression has been detected in myasthenia gravis. Abnormally wide spread expressions in mammary gland and colon have also been reported in breast and colonrectal carcinomas. Although it shows high amino acid identity with cathepsin L (Cat. No. 219382), these two cathepsins have very different tissue distribution and substrate specificity. As a result of glycosylation, this recombinant enzyme migrates ~40 kDa on SDS-PAGE under either reducing or non-reducing conditions. Requires activation prior to use.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will cleave 1 pmol the fluorescent substrate Z-LR-AMC per min at 25°C, pH 5.5.

Physical form

In 800 mM NaCl, 25 mM sodium acetate, 0.8 M NaCl, pH 5.0.

Reconstitution

To activate rhCathepsin V, pre-incubate it at 20 µg/ml in 25 mM NaOAc, 100 mM NaCl, 5 mM DTT, pH 5.5 at room temperature. Following initial thaw aliquot and freeze (-70°C).

Other Notes

Tolosa, E., et al. 2003. J. Clin. Invest.112, 517.
Turk, V., et al. 2001. EMBO J.20, 4629.
Somoza, J.R., et al. 2000. Biochemistry39, 12543.
Bromme, D., et al. 1999. Biochemistry38, 2377.
Adachi, W., et al. 1998. Invest. Ophthalmol. Vis. Sci.39, 1789.
Santamaria, I., et al. 1998. Cancer Res.58, 1624.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service