Skip to Content
Merck
All Photos(1)

Key Documents

203729

Sigma-Aldrich

(+)-Brefeldin A, Eupenicillium brefeldianum

Specifically and reversibly blocks translocation of proteins from the endoplasmic reticulum (ER) to the Golgi apparatus without affecting endocytosis or lysosome function.

Synonym(s):

(+)-Brefeldin A, Eupenicillium brefeldianum, 1,6,7,8,9,11a,12,13,14,14a-Decahydro-1,13-dihydroxy-6-methyl-4H-cyclopent[f]oxacyclotridecin-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24O4
CAS Number:
Molecular Weight:
280.36
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

methanol: 2 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

O1[C@H](CCC\C=C\[C@H]2[C@H]([C@H](\C=C\C1=O)O)C[C@H](C2)O)C

InChI

1S/C16H24O4/c1-11-5-3-2-4-6-12-9-13(17)10-14(12)15(18)7-8-16(19)20-11/h4,6-8,11-15,17-18H,2-3,5,9-10H2,1H3/b6-4+,8-7+/t11-,12+,13-,14+,15-/m0/s1

InChI key

KQNZDYYTLMIZCT-PNFJWZTBSA-N

General description

Specifically and reversibly blocks translocation of proteins from the endoplasmic reticulum (ER) to the Golgi apparatus without affecting endocytosis or lysosome function. Inhibitor of HDL-mediated cholesterol efflux. Causes disassembly of the Golgi complex and ER swelling in a variety of mammalian cell lines at <40 ng/ml. Blocks binding of ADP-ribosylation factor to the Golgi and inhibits the GDP-GTP exchange. Inhibits the activity of BIG1 and BIG2, the guanine-nucleotide exchange proteins for ADP-ribosylation factors. Also available as a 25 mM solution in DMSO (Cat. No. 500583).

Biochem/physiol Actions

Cell permeable: no
Primary Target
Blocks translocation of proteins from the endoplasmic reticulum (ER) to the Golgi apparatus
Product does not compete with ATP.
Reversible: yes

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for 2 months at -20°C.

Other Notes

Yamagi, R., et al. 2000. Proc. Natl. Acad. Sci. USA 97, 2567.
Moriango, N., et al. 1996. Proc. Natl. Acad. Sci. USA 93, 12856.
Mendez, A.J. 1995. J. Biol. Chem. 270, 5891.
Ridgway, N.D., et al. 1995. J. Biol. Chem. 270, 8023.
Tsai, S.C., et al. 1994. Proc. Natl. Acad. Sci. USA91, 3063.
Ktistakis, N.Y., et al. 1992. Nature356, 344.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tonke K Raaijmakers et al.
Journal for immunotherapy of cancer, 8(1) (2020-05-29)
Tumor ablation techniques, like cryoablation, are successfully used in the clinic to treat tumors. The tumor debris remaining in situ after ablation is a major antigen depot, including neoantigens, which are presented by dendritic cells (DCs) in the draining lymph
Shreyasi Maity et al.
Infection and immunity, 91(10), e0026723-e0026723 (2023-09-19)
In view of the antagonism of Wnt5A signaling toward microbial pathogens, we were interested in evaluating the therapeutic potential of recombinant Wnt5A (rWnt5A) in curbing Leishmania donovani infection. Initially, using L. donovani-infected RAW 264.7 and peritoneal macrophages, we demonstrated that
Daniela Cunha et al.
Cells, 13(11) (2024-06-19)
Basal cell carcinomas (BCCs) and squamous cell carcinomas (SCCs) are high-incidence, non-melanoma skin cancers (NMSCs). The success of immune-targeted therapies in advanced NMSCs led us to anticipate that NMSCs harbored significant populations of tumor-infiltrating lymphocytes with potential anti-tumor activity. The
Yan Lin et al.
Journal of immunology research, 2021, 8899674-8899674 (2021-01-26)
Tuberculosis is the world's most deadly infectious disease, with 10 million people falling ill and 1.5 million people dying from the disease every year. With the increasing number of drug-resistant Mycobacterium tuberculosis (MTB) strains and prevalence of coinfection of MTB
Meirong Li et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 129, 110425-110425 (2020-06-23)
Acute leukemia is a common hematological malignancy. Despite recent promising progress, the prognosis of acute leukemia patients remains to be improved. New therapies are therefore still needed. Salmonella typhimurium has been shown to be highly effective as an anti-tumor agent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service