Skip to Content
Merck
All Photos(2)

Key Documents

17-601

Sigma-Aldrich

ChIPAb+ Sp1 - ChIP Validated Antibody and Primer Set

from rabbit

Synonym(s):

Transcription factor Sp1, TSFP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

clone

polyclonal

species reactivity

rat, human, mouse

manufacturer/tradename

ChIPAb+
Upstate®

technique(s)

ChIP: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

Every lot of the ChIPAb+ line of antibodies is individually validated for chromatin precipitation, in order to guarantee successful ChIP assays every time. Each antibody includes a control primer set for performance confirmation. Sp1 antibody and the negative control antibody (rabbit normal IgG) can be used to demonstrate that the Sp1 antibody is functionally validated in the precipitation of Sp1 associated chromatin.
The qPCR primers included flank the Sp1 binding site in human DHFR promoter.
Transcription factor SP1 can activate or repress transcription in response to physiological and pathological stimuli. Binds with high affinity to GC-rich motifs and regulates the expression of a large number of genes involved in a variety of processes such as cell growth, apoptosis, differentiation and immune responses. Highly regulated by post-translational modifications (phosphorylations, sumoylation, proteolytic cleavage, glycosylation and acetylation). Binds also the PDGFR-alpha G-box promoter. May have a role in modulating the cellular response to DNA damage. Implicated in chromatin remodeling. Plays a role in the recruitment of SMARCA4/BRG1 on the c-FOS promoter. Plays an essential role in the regulation of FE65 gene expression. In complex with ATF7IP, maintains telomerase activity in cancer cells by inducing TERT and TERC gene expression. Isoform 3 is a stronger activator of transcription than isoform 1. Positively regulates the transcription of the core clock component ARNTL/BMAL1.

Specificity

Sp1

Immunogen

Full length human Sp1 protein

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
SP1 ChIP validated antibody & primer set including the ChIP-grade antibody & the specific control PCR primers used for chromatin immunoprecipitation of SP1.
Western Blot Analysis:
HeLa nuclear extract was resolved by electrophoresis, transferred to nitrocellulose and probed with anti-Sp1 (0.2 μg/mL). Proteins were visualized using a goat anti-rabbit secondary antibody conjugated to HRP and a chemiluminescence detection system (Please see figures).

Packaging

25 assays per kit, ~4μg per chromatin immunoprecipitation.

Quality

Chromatin immunoprecipitation:
Sonicated chromatin prepared from 1 x 106 HeLa cells was subjected to chromatin immunoprecipitation using 4 μg of either the negative control antibody, Rabbit Normal IgG or Rabbit Anti-Sp1 and the Magna ChIPTM A kit (Cat.# 17-610) Rapid Protocol (2 hour IP). Successful enrichment of Sp1 associated DNA fragments was verified by qPCR using ChIP Primers DHFR flanking the human DHFR promoter that contains a Sp1 binding site (Please see figures).
Please refer to the EZ-Magna ChIP A (Cat.# 17-408)
or EZ-ChIP (Cat.# 17-371) kit protocols for
experimental details.

Target description

105 kDa and 95 kDa

Physical form

Anti-Sp1 (Rabbit polyclonal IgG). 1 vial containing 100 μg of protein A purified IgG in 100 μL of 0.014 M phosphate buffer, pH 7.6, 0.175 M NaCl, 0.07% sodium azide and 30% glycerol. Store at -20°C. The Sp1 antibody is made against full length human Sp1 protein and can recognize Sp1, MW 105 kDa and 95 kDa, of human, mouse and rat origins.

Normal Rabbit IgG. One vial containing 125 μg of normal rabbit IgG in 125 μL. Store at -20°C.

ChIP primers DHFR. 1 vial containing 75 μL of 5 μM of each control primer specific for human dihydrofolate reductase (DHFR) promoter. Store at -20°C.
FOR: TCG CCT GCA CAA ATA GGG AC
REV: AGA ACG CGC GGT CAA GTT T
Format: Purified
Protein A purified

Storage and Stability

1 year at -20°C from date of shipment.

Analysis Note

Control
Includes negative control rabbit IgG and control primers specific for human dihydrofolate reductase (DHFR) promoter.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silvia Salatino et al.
Molecular endocrinology (Baltimore, Md.), 30(7), 809-825 (2016-05-18)
The peroxisome proliferator-activated receptor-γ coactivator 1α (PGC-1α) coordinates the transcriptional network response to promote an improved endurance capacity in skeletal muscle, eg, by coactivating the estrogen-related receptor-α (ERRα) in the regulation of oxidative substrate metabolism. Despite a close functional relationship
Sylvain Fauquenoy et al.
Scientific reports, 7, 43221-43221 (2017-03-04)
Human T-lymphotropic Virus type 1 (HTLV-1) infection is characterized by viral latency in the majority of infected cells and by the absence of viremia. These features are thought to be due to the repression of viral sense transcription in vivo.
Yuanyuan Zhao et al.
Molecular oncology, 12(3), 305-321 (2017-12-08)
Transforming growth factor-beta (TGF-β) functions as a potent proliferation inhibitor and apoptosis inducer in the early stages of breast cancer, yet promotes cancer aggressiveness in the advanced stages. The dual effect of TGF-β on cancer development is known as TGF-β
Identification and characterization of the human NOL7 gene promoter.
Mankame, TP; Zhou, G; Lingen, MW
Gene null
Paulisally Hau Yi Lo et al.
PloS one, 8(4), e61279-e61279 (2013-04-18)
Hepatitis C virus (HCV) infection is the major cause of hepatocellular carcinoma (HCC) in Japan. We previously identified the association of SNP rs2596542 in the 5' flanking region of the MHC class I polypeptide-related sequence A (MICA) gene with the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service