Skip to Content
Merck
All Photos(1)

Key Documents

1290-OP

Millipore

Acrylamide/Bis-acrylamide

OmniPur® Grade, 40% solution, 19:1 ratio, Suitable for DNA and RNA separations and sequencing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105319

Quality Level

form

solution

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clear colorless

shipped in

wet ice

storage temp.

2-8°C

General description

Suitable DNA and RNA separations and sequencing. Molecular biology grade. Reconstitute with deionized water to desired final concentration. Premixed solutions offer standard ratios of acrylamide to bis-acrylamide for convenience and safety. The hazard of handling powdered acrylamide is eliminated.

Specifications:
DNase: None detected
Protease: None detected
RNase: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabienne C Fiesel et al.
Bio-protocol, 6(19) (2017-06-03)
This protocol describes the in vitro phosphorylation of ubiquitin and Parkin by the kinase PINK1 using recombinant proteins. Both substrates, ubiquitin and Parkin, are phosphorylated at the conserved serine 65 residue (pS65-ubiquitin and pS65-Parkin). The protocol also includes the use

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service