Skip to Content
Merck
All Photos(1)

Key Documents

1.16989

Supelco

Formaldehyde Test Strips, reflectometric

1.0-45.0 mg/L (HCHO), REFLECTOQUANT®

Synonym(s):

Formaldehyde chemical test strips, Formaldehyde test strip for instrument reader

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Formaldehyde Test, reflectometric, 1.0-45.0 mg/L (HCHO), for use with REFLECTOQUANT®

description

quantitative

Quality Level

product line

Reflectoquant®

specific analyte(s)

formaldehyde

measuring range

1.0-45.0 mg/L (HCHO)

compatibility

RQflex® 10 Plus
RQflex® 10
RQflex® 20
for use with REFLECTOQUANT®

detection method

reflectometric

storage temp.

15-25°C

General description

Method: reflectometric with test strips and reagent 1.0 - 45.0 mg/l HCHO Reflectoquant®
The Reflectoquant Formaldehyde test includes test strips and all necessary additional reagents. Formaldehyde is used as a disinfectant for surfaces, medical equipment and also as a raw material in binding agents, fixing agents or as a preservative. Formaldehyde reacts with a Triazole to form a purple-red tetrazine that is determined reflectometrically on the RQflex. For pre-testing a sample in an economic and quick way to estimate the formaldehyde concentration and a possible dilution factor prior to using this quantitative method we recommend our semi-quantitative MQuant test strips for formaldehyde. Further quantitative determination methods can be found with our Spectroquant tests.

Analysis Note

function control: conforms
Assay
- standard 1:
4.5 - 5.5 mg/l
- standard 2: 13.5 - 16.5 mg/l
- standard 3: 27.0 - 33.0 mg/l

Legal Information

REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Preparation of a standard solution for Formaldehyde

Preparation of a standard solution for Formaldehyde

Preparation of a standard solution for Formaldehyde

Preparation of a standard solution for Formaldehyde

Related Content

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service