Skip to Content
Merck
All Photos(2)

Key Documents

1.11972

Millipore

GranuCult® Tryptose sulfite cycloserine agar (base)

ISO 7937, ISO 14189, APHA, suitable for microbiology

Synonym(s):

TSC agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

APHA
ISO 14189
ISO 7937

Quality Level

sterility

non-sterile

form

granular

manufacturer/tradename

GranuCult® prime

technique(s)

microbiological culture: suitable

pH

7.4-7.8 (25 °C in H2O)

solubility

42 g/L

bulk density

660 kg/m3

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Clostridium spp.

General description

GranuCult® Tryptose sulfite cycloserine agar (base) is intended for the isolation and differentiation of Clostridium perfringens. The superior nutrient base provides optimal conditions for the development of Clostridia. Colonies producing hydrogen sulfide are characterized by blackening due to the reaction with sulfite and iron salt. Cycloserine inhibits the accompanying bacterial flora and may cause the colonies that develop to remain smaller. It also reduces diffusion and thus the disturbing blackening around the Clostridium perfringens colonies. TSC agar with MUP contains a fluorogenic substrate for the alkaline and acid phosphatase in addition to cycloserine 4-Methylumbelliferylphosphate (MUP). The acid phosphatase is a high specific indicator for Clostridium perfringens. It splits the fluorogenic substrate MUP, forming 4-methylumbelliferone, which can be identified by its fluorescence in long wave UV light. Thus, a strong suggestion for the presence of Clostridium perfringens can be obtained.

Application

GranuCult® Tryptose sulfite cycloserine agar (base) is intended for the isolation and differentiation of Clostridium perfringens from food and animal feed, water and other materials. TSC agar with MUP can be prepared using Clostridium perfringens Supplement (1.00888) to detect acid phosphatase by its fluorescence under UV light.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vasileios Tsiouris et al.
Foods (Basel, Switzerland), 9(5) (2020-05-10)
Whey is a highly nutritious byproduct of the cheese industry that can be used effectively in the animal feed industry. However, the use of whey in poultry diets is limited by its high lactose and mineral contents. The objective of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service