Skip to Content
Merck
All Photos(1)

Key Documents

1.01711

Sigma-Aldrich

Barium carbonate

Technipur®

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaCO3
CAS Number:
Molecular Weight:
197.34
Beilstein:
7045119
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21

Quality Level

pH

7-8 (16 °C, .016 g/L in H2O)

mp

>1450 °C (decomposition)

SMILES string

[Ba++].[O-]C([O-])=O

InChI

1S/CH2O3.Ba/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

AYJRCSIUFZENHW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Barium Carbonate (BaCO3) is one of the commercially important barium compounds. It is widely used in ceramics, paints, enamels, plastics, etc. It is particularly of interest in dental and lighting applications.

Application

Barium carbonate (BaCO3) can be used as a starting material to prepare:
  • SiO2-BaO composites for use as inorganic filler materials in dental composite resins
  • silicate-containing ceramic glasses for dental applications
  • commercial phosphors for solid-state lighting (SSL) applications, especially light-emitting diodes (LEDs)
It can also be used as a cation source for isovalent substitution in yellow phosphor hosts employed in solid-state white lighting applications.

Legal Information

Technipur is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of aqueous sol-gel method for synthesis of nanostructured SiO2-BaO powder for use in dental composite resins
Ghosh NN
Ceramic Nanomaterials and Nanotechnologies, 207-212 (2012)
Synthesis of SiO2-BaO powder by aqueous sol-gel processing for use in dental composite resins
Ghosh NN and Pramanik P
British Ceramic Transactions, 95(6), 267-270 (1996)
Hydrated Behaviors and Mechanical Properties of SilicateContaining Dental Glass Ceramics
Sheu TS, et al.
Proceedings of the National Science Council, Republic of China. Part B, Life Sciences, 24(4), 301-305 (2000)
Chapter 61?inorganic and organometal pesticides
Clarkson TW, et al.
Handbook of Pesticide Toxicology, Two-Volume Set: Principles and Agents, Volume 1 (2001)
Sr2.975-xBaxCe0.025AlO4F: a Highly Efficient Green-Emitting Oxyfluoride Phosphor for Solid State White Lighting
Im WB, et al.
Chemistry of Materials, 22(9), 2842?2849-2842?2849 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service