Skip to Content
Merck
All Photos(1)

Key Documents

1.01567

Supelco

Sodium hydroxide on support

granulated about 0.8-1.6 mm for elementary analysis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352305
NACRES:
NA.21

Quality Level

form

solid

pH

13.8 (20 °C, 50 g/L in H2O)

bulk density

800 kg/m3

storage temp.

2-30°C

Analysis Note

Absorption capacity for CO₂: ≥ 50 %
Loss on ignition (800 °C): ≤ 25 %
Bulk density: about 70 g/100 ml

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service