Skip to Content
Merck
All Photos(1)

Key Documents

06-991

Sigma-Aldrich

Anti-PKCε Antibody

Upstate®, from rabbit

Synonym(s):

protein kinase C, epsilon

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human, mouse

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PRKCE(5581)

General description

Members of the protein kinase C family fall into three categories: conventional (PKC alpha, beta I, beta II, gamma), novel (delta, epsilon, eta, mu, theta), and atypical (lambda, zeta). Conventional PKC isoforms are dependent on calcium and diacylglycerol (DAG) for their activity, whereas novel PKC isoforms are calcium dependent, and atypical PKC isoforms are calcium and DAG independent.

Specificity

Predicted to cross-react with rabbit.
Recognizes PKCε, Mr 95 kDa.

Immunogen

KLH-conjugated, synthetic peptide corresponding to the C-terminal variable (V5) region, amino acids 726-737 (KGFSYFGEDLMP), of mouse Protein Kinase C Epsilon (PKCε). The immunizing sequence is identical in human, mouse, rat and rabbit.

Application

Detect PKCε using this Anti-PKCε Antibody validated for use in IH, IP & WB.
Immunoprecipitation:
An independent lab has reported this antibody immunoprecipitates PKCε.

Immunohistochemistry:
An independent lab has reported this antibody detects PKCε.
Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling

Quality

Routinely evaluated by Western Blot on Jurkat cell lysate.

Western Blot Analysis:
0.5-2 μg/mL of this lot detected PKCε in Jurkat cell lysate. 0.5-2 μg/mL of a previous lot detected PKCε in a RIPA lysate from Jurkat cells, mouse brain extract and rat brain extract.

Target description

Approx. 95

Physical form

Format: Purified
Protein A purified
Purified rabbit IgG in buffer containing 0.014 M phosphate buffer, pH 7.6, 0.175 M NaCl, 0.07% sodium azide, and 30% glycerol.
Store at -20°C.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
Positive Antigen Control: Catalog #12-303, Jurkat cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Overexpression of protein kinase C-delta and -epsilon in NIH 3T3 cells induces opposite effects on growth, morphology, anchorage dependence, and tumorigenicity
Mischak, H., et al
The Journal of Biological Chemistry, 268, 6090-6096 (1993)
François Bordeleau et al.
Molecular biology of the cell, 21(10), 1698-1713 (2010-04-02)
Keratins are intermediate filament (IF) proteins of epithelial cells, expressed as pairs in a lineage/differentiation manner. Hepatocyte and hepatoma cell IFs are made solely of keratins 8/18 (K8/K18), the hallmark of all simple epithelia. Cell attachment/spreading (adhesion) and migration involve
Expression of four protein kinase C isoforms in rat fibroblasts. Differential alterations in ras-, src-, and fos-transformed cells
Borner, C., et al
The Journal of Biological Chemistry, 267, 12900-12910 (1992)
Andressa G Amaral et al.
Pancreas, 39(6), 847-855 (2010-08-11)
We investigated the influence of leucine supplementation on insulin secretion and on some proteins related to insulin secretion in malnourished mice. Swiss mice (aged 21 days) received isocaloric normo-17% (NP) or 6% low-protein (LP) diet for 120 days. Half of
Post-ischaemic activation of kinases in the pre-conditioning-like cardioprotective effect of the platelet-activating factor.
Penna C, Mognetti B, Tullio F, Gattullo D, Mancardi D, Moro F, Pagliaro P, Alloatti G
Acta Physiologica (Oxford, England) null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service