Skip to Content
Merck
All Photos(1)

Key Documents

05-565

Sigma-Aldrich

Anti-Nucleolin Antibody, clone 3G4B2

culture supernatant, clone 3G4B2, Upstate®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

3G4B2, monoclonal

species reactivity

human, canine, monkey

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NCL(4691)

General description

Nucleolin, also known as C23, is the major nucleolar protein of exponentially growing eukaryotic cells. It is a ubiquitous conserved multi-domain phosphoprotein found in all vertebrate species. Nucleolin contains a specific bipartite nuclear localization signal sequence and three different structural domains. Nucleolin is a multifunctional protein involved in most steps of ribosome biogenesis. Nucleolin has been implicated in chromatin structure, rDNA transcription, rRNA maturation, ribosome assembly, cytokinesis, nucleogenesis, cell proliferation and growth, and
nucleo-cytoplasmic transport. Nucleolin also exhibits autodegradation, DNA and RNA helicase activities, and DNA-dependent ATPase activity. Nucleolin activities may be regulated by proteolysis, methylation, ADPribosylation, and phosphorylation by several kinases, including casein kinase II (CK2), p34cdc2, and protein kinase C-ζ.

Specificity

nucleolin

Immunogen

Nuclear matrix extract from MDCK cells

Application

Anti-Nucleolin Antibody, clone 3G4B2 is a high quality Mouse Monoclonal Antibody for the detection of Nucleolin & has been validated in IP, WB, ICC.

Quality

routinely evaluated by immunoblot on HeLa nuclear extract

Target description

100kDa

Linkage

Replaces: MABE466

Physical form

mouse culture supernatant containing 0.05% sodium azide prior to the addition of glycerol to 30%

Analysis Note

Control
Positive Antigen Control: Catalog #12-309, Hela cell nuclear extract. Add an equal volume of Laemmli reducing sample buffer to 10 μL of extract and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced extract per lane for minigels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ingrid Morland et al.
Nucleic acids research, 30(22), 4926-4936 (2002-11-16)
The mild phenotype associated with targeted disruption of the mouse OGG1 and NTH1 genes has been attributed to the existence of back-up activities and/or alternative pathways for the removal of oxidised DNA bases. We have characterised two new genes in
Dominique Legrand et al.
European journal of biochemistry, 271(2), 303-317 (2004-01-14)
Lactoferrin (Lf), a multifunctional molecule present in mammalian secretions and blood, plays important roles in host defense and cancer. Indeed, Lf has been reported to inhibit the proliferation of cancerous mammary gland epithelial cells and manifest a potent antiviral activity
The PI3K/Akt/mTOR pathway is implicated in the premature senescence of primary human endothelial cells exposed to chronic radiation.
Yentrapalli, R; Azimzadeh, O; Sriharshan, A; Malinowsky, K; Merl, J; Wojcik et al.
Testing null
Tatiana A Fedotcheva et al.
Pharmaceutics, 13(12) (2021-12-29)
Agrimoniin is a polyphenol from the group of tannins with antioxidant and anticancer activities. It is assumed that the anticancer action of agrimoniin is associated with the activation of mitochondria-dependent apoptosis, but its mitochondrial targets have not been estimated. We
Malika Hamdane et al.
The Journal of biological chemistry, 278(36), 34026-34034 (2003-06-27)
Among tau phosphorylation sites, some phosphoepitopes referred to as abnormal ones are exclusively found on tau aggregated into filaments in Alzheimer's disease. Recent data suggested that molecular mechanisms similar to those encountered during mitosis may play a role in abnormal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service