Skip to Content
Merck
All Photos(1)

Key Documents

V-067

Supelco

(−)-Riboflavin

100 μg/mL (1% ammonium acetate in 50:50 methanol:water), certified reference material, ampule of 1 mL, Cerilliant®

Synonym(s):

Lactoflavin, Vitamin B2, Vitamin G

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N4O6
CAS Number:
Molecular Weight:
376.36
Beilstein:
07825
UNSPSC Code:
12352205
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL (1% ammonium acetate in 50:50 methanol:water)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

mp

290 °C (dec.)

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

O[C@H]([C@H](O)[C@H](O)CO)CN1C2=CC(C)=C(C)C=C2N=C3C1=NC(NC3=O)=O

InChI

1S/C17H20N4O6/c1-7-3-9-10(4-8(7)2)21(5-11(23)14(25)12(24)6-22)15-13(18-9)16(26)20-17(27)19-15/h3-4,11-12,14,22-25H,5-6H2,1-2H3,(H,20,26,27)/t11-,12+,14-/m0/s1

InChI key

AUNGANRZJHBGPY-SCRDCRAPSA-N

General description

Riboflavin is an essential B vitamin for maintaining human health with key roles in energy metabolism as well as a critical component of enzyme cofactors and flavoproteins.

Application

Riboflavin (vitamin B2) may be used as a certified reference material (CRM) for the determination of the analyte in foodstuffs, biological samples and pharmaceuticals by different chromatography techniques.
This certified solution standard is suitable for use as starting materials in the preparation of calibrators, controls, or linearity standards in a variety of testing applications from diagnostic testing of vitamin B2 deficiency by HPLC to quality control testing of vitamin B2-based dietary supplements, fortified foods or pharmaceutical preparations.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 3 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

75.2 °F

Flash Point(C)

24 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chromatographic determination of riboflavin and its derivatives in food
Gliszczynska-Swiglo A and Koziolowa A
Journal of Chromatography A, 881(1-2), 285-297 (2000)
Determination of thiamine, riboflavin and pyridoxine in pharmaceuticals by synchronous fluorescence spectrometry in organized media
Garc?a L, et al.
Analytica Chimica Acta, 434(2), 193-199 (2001)
Determination of riboflavin in urine by capillary electrophoresis--blue light emitting diode-induced fluorescence detection combined with a stacking technique
Su An-Kai and Lin Cheng-Huang
Journal of Chromatography. B, Biomedical Sciences and Applications, 785(1), 39-46 (2003)
Rapid determination of thiamine, riboflavin, niacinamide, pantothenic acid, pyridoxine, folic acid and ascorbic acid in Vitamins with Minerals Tablets by high-performance liquid chromatography with diode array detector
Jin P, et al.
Journal of Pharmaceutical and Biomedical Analysis, 70(2), 151-157 (2012)
Rapid determination of vitamin B2 (riboflavin) in plasma by HPLC
Petteys BJ and Frank Elizabeth L
Clinica Chimica Acta; International Journal of Clinical Chemistry, 412(1-2), 38-43 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service