Skip to Content
Merck
All Photos(1)

Key Documents

T-039

Supelco

Topiramate solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H21NO8S
CAS Number:
Molecular Weight:
339.36
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CC1(C)O[C@@H]2CO[C@@]3(COS(N)(=O)=O)OC(C)(C)O[C@H]3[C@@H]2O1

InChI

1S/C12H21NO8S/c1-10(2)18-7-5-16-12(6-17-22(13,14)15)9(8(7)19-10)20-11(3,4)21-12/h7-9H,5-6H2,1-4H3,(H2,13,14,15)/t7-,8-,9+,12+/m1/s1

InChI key

KJADKKWYZYXHBB-XBWDGYHZSA-N

General description

A certified solution standard suitable for use in LC/MS or GC/MS applications for pharmaceutical research, clinical toxicology, or forensic testing.  Topiramate, sold under trade name Topamax®, is an antiepileptic drug used for treatment of seizures. Topiramate in combination with the stimulant phentermine constitute the active compounds in the new weight lose drug Qsymia®.

Application


  • Topiramate research solution: Research focuses on the development of intravenous formulations of topiramate for rapid intervention in seizure emergencies, highlighting its potential in emergency medicine and critical care (Epilepsy Behav, 2023; doi: 10.1016/j.yebeh.2023.109158).

  • Topiramate liquid formulation: Innovations in drug delivery include the use of topiramate-chitosan nanoparticles, which offer enhanced drug stability and targeted delivery, particularly in addiction treatment settings (Neurochem Int, 2021; doi: 10.1016/j.neuint.2021.105157).

  • Anticonvulsant biochemical reagent: Topiramate′s role as an anticonvulsant is further explored in studies focusing on its pharmacokinetic properties and interaction with other medications, emphasizing its importance in personalized medicine (Ther Drug Monit, 2019; doi: 10.1097/FTD.0000000000000600).

  • Topiramate in vitro study: The compound is utilized in in vitro setups to investigate its effects on neuronal excitability, providing insights into its mechanism of action and potential therapeutic applications (Epilepsia, 2019; doi: 10.1111/epi.16073).

  • Topiramate pharmacological research: Studies include the development of advanced analytical methods for the quantification of topiramate in biological samples, supporting its clinical monitoring and therapeutic drug management (J Pharm Biomed Anal, 2015; doi: 10.1016/j.jpba.2015.01.035).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Qsymia is a registered trademark of Vivus, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Topamax is a registered trademark of Johnson & Johnson

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J M Riffitts et al.
Journal of pharmaceutical and biomedical analysis, 19(3-4), 363-371 (2000-03-07)
An accurate and robust method involving liquid liquid extraction and capillary gas chromatographic (GC) assay with nitrogen phosphorus detection (NPD) was developed and validated for the quantitative determination of topiramate [2,3:4,5-bis-O-(-1-methylethylidene)-beta-D-fructopyranose sulfamate], Topamax, an anticonvulsant drug, in human plasma, urine
Steve S Chung et al.
Epilepsia, 55(7), 1077-1087 (2014-06-07)
To evaluate the efficacy and safety of USL255, Qudexy(™) XR (topiramate) extended-release capsules, as an adjunctive treatment for refractory partial-onset seizures (POS) in adults taking one to three concomitant antiepileptic drugs. In this global phase III study (PREVAIL; NCT01142193), 249
Annie M Clark et al.
Epilepsia, 55(7), 1069-1076 (2014-05-28)
Evaluate the pharmacokinetics (PK), safety, and tolerability of single doses of once-daily USL255, Qudexy XR (topiramate) extended-release capsules, over a wide dosing range. Two single-dose, phase I studies in healthy adults were used to evaluate the PK profile and maximum
Elizabeth M Kelly et al.
Journal of managed care pharmacy : JMCP, 19(8), 642-654 (2013-10-01)
Obesity may lead to the development of multiple chronic disease states, including hypertension, dyslipidemia, and type 2 diabetes mellitus. Over a half billion adults worldwide are affected by obesity, and more than two-thirds of adults are either obese or overweight
William Timothy Garvey
Expert opinion on drug safety, 12(5), 741-756 (2013-06-07)
Weight-management options include lifestyle modifications, bariatric surgery and, until recently, limited pharmacotherapy. Phentermine and topiramate extended-release (phentermine/topiramate ER) has recently been approved in the USA for chronic weight management in obese adults and overweight adults with weight-related co-morbidities in conjunction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service