Skip to Content
Merck
All Photos(1)

Documents

T-030

Supelco

Trazodone hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23Cl2N5O
CAS Number:
Molecular Weight:
408.32
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

Cl.Clc1cccc(c1)N2CCN(CCCN3N=C4C=CC=CN4C3=O)CC2

InChI

1S/C19H22ClN5O.ClH/c20-16-5-3-6-17(15-16)23-13-11-22(12-14-23)8-4-10-25-19(26)24-9-2-1-7-18(24)21-25;/h1-3,5-7,9,15H,4,8,10-14H2;1H

InChI key

OHHDIOKRWWOXMT-UHFFFAOYSA-N

Gene Information

General description

Trazodone is a antidepressant drug sold under trade names such as Desyrel® and Oleptro® for treatment of anxiety disorder, depression, and insomnia. Trazadone, a serotonin antagonist and reuptake inhibitor (SARI), also has anti-anxiety and hypnotic effects. This certified Snap-N-Spike® solution is suitable for use in clinical toxicology, forensic analysis, or urine drug testing methods by LC-MS/MS or GC/MS.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Desyrel is a registered trademark of Aziende Chimiche Riunite Angelini Francesco (A.C.R.A.F.) S.p.A.
Oleptro is a registered trademark of Angelini Pharma Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Georgotas et al.
Pharmacotherapy, 2(5), 255-265 (1982-09-01)
Trazodone is a new antidepressant agent that was recently introduced in the United States. It has a unique pharmacological profile that is not typical of either tricyclic or monoamine oxidase inhibitor antidepressants. As such it represents a new class of
Andrea Fagiolini et al.
CNS drugs, 26(12), 1033-1049 (2012-11-30)
Trazodone is a triazolopyridine derivative that belongs to the class of serotonin receptor antagonists and reuptake inhibitors (SARIs). The drug is approved and marketed in several countries worldwide for the treatment of major depressive disorder (MDD) in adult patients. In
Marek Jarema et al.
Psychiatria polska, 45(4), 611-625 (2012-01-12)
The pharmacological properties and possible clinical use of trazodon in the treatment of depression are presented. Trazodon is the only antidepressant from the SARI (Serotonin Antagonists and Reuptake Inhibitors) group available in Poland. It has a wide pharmacodynamic profile (being
Arnaud Devresse et al.
Acta gastro-enterologica Belgica, 75(2), 263-265 (2012-08-09)
We present the case of a 26-year-old man with schizoid personality disorder who suffered from a very focal and transparietal necrosis of the sigmoid after an overdose of atypical neuroleptics. This is a singular, rather unknown and potentially lethal side
Ramez Ghanbari et al.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology, 22(7), 518-526 (2011-12-14)
Previous study has documented the long-term effects of the antidepressant trazodone on the serotonin (5-HT) system. The present work examined the impact of sustained trazodone on ventral tegmental area (VTA) dopamine (DA) and locus ceruleus (LC) norepinephrine (NE) neurons firing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service