Skip to Content
Merck
All Photos(1)

Key Documents

P-066S

Supelco

Pregabalin solution

new

1 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Synonym(s):

Pregabalin solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17NO2
CAS Number:
Molecular Weight:
159.23
MDL number:
UNSPSC Code:
12352200

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol

format

single component solution

storage temp.

-10 to -25°C

SMILES string

CC(CC([H])(CN)CC(O)=O)C

InChI

1S/C8H17NO2/c1-6(2)3-7(5-9)4-8(10)11/h6-7H,3-5,9H2,1-2H3,(H,10,11)

InChI key

AYXYPKUFHZROOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sold under the trade name Lyrica®, pregabalin is an antiepileptic drug used for treatment of seizures, fibromyalgia, diabetic peripheral neuropathy, generalized anxiety disorder, and chronic neuropathic pain. This certified reference standard is suitable for pregabalin GC/MS or LC/MS applications from forensic analysis, clinical toxicology and pain prescription monitoring to urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Lyrica is a registered trademark of C.P. Pharmaceuticals International C.V.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service