Skip to Content
Merck
All Photos(1)

Key Documents

O-023

Supelco

Oxazepam glucuronide solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H19ClN2O8
CAS Number:
Molecular Weight:
462.84
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

O[C@@H]1[C@@H](O)[C@H](OC2N=C(c3ccccc3)c4cc(Cl)ccc4NC2=O)O[C@@H]([C@H]1O)C(O)=O

InChI

1S/C21H19ClN2O8/c22-10-6-7-12-11(8-10)13(9-4-2-1-3-5-9)24-19(18(28)23-12)32-21-16(27)14(25)15(26)17(31-21)20(29)30/h1-8,14-17,19,21,25-27H,(H,23,28)(H,29,30)/t14-,15-,16+,17-,19?,21-/m0/s1

InChI key

FIKQKGFUBZQEBL-IFBJMGMISA-N

General description

Oxazepam glucuronide is a major urinary metabolite of the benzodiazepine drug, oxazepam. Oxazepam, marketed as Alepam, Medopam, or Murelax®, has been used extensively since the 1960s for treatment of anxiety and insomnia and in the control of symptoms of alcohol withdrawal. This certified reference solution is suitable for GC/MS or LC/MS applications with oxazepam or oxazepam glucuronide such as urine drug testing, pain prescription monitoring, toxicology analysis, or forensic testing.

Application


  • Advanced Analytical Techniques for Benzodiazepine Metabolites: Oxazepam glucuronide has been targeted in studies using high-performance liquid chromatography with tandem mass spectrometry (HPLC-MS/MS). This method allows for the sensitive and automatic determination of diazepam and its metabolites, including oxazepam glucuronide, in human oral fluid, highlighting its utility in forensic and clinical toxicology (Jiang et al., 2016).

  • Forensic Applications: Oxazepam glucuronide is directly determined using μElution solid-phase extraction and liquid chromatography-tandem mass spectrometry (LC-MS/MS) in human whole blood. This technique is crucial for forensic science, providing reliable data for legal and medical investigations involving benzodiazepine use (Wang et al., 2013).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Murelax is a registered trademark of Aspen Pharma Pty Ltd
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E J Pantuck et al.
Clinical pharmacology and therapeutics, 35(2), 161-169 (1984-02-01)
Ten healthy subjects were fed three diets for 10 days each: a control diet, a cabbage and brussels sprouts--containing diet, and the control diet a second time. Oxazepam was taken on day 7 and acetaminophen on day 10 of each
M Patel et al.
Pharmacogenetics, 5(1), 43-49 (1995-02-01)
1,4-Benzodiazepine anxiolytics such as diazepam and halazepam are converted in vivo to oxazepam, an active metabolite with a hydroxyl group at the asymmetric C3 position. D-glucuronic acid couples with the C3 hydroxyl group of oxazepam to form pharmacologically inactive diastereomeric
S F Sisenwine et al.
Drug metabolism and disposition: the biological fate of chemicals, 14(1), 41-45 (1986-01-01)
The metabolic fate of oxazepam in the rat is more complex than in larger animal species. Condensation of the diazepinyl ring and phase 1 transformations which lead presumedly via an epoxide to metabolites hydroxylated on the 5-phenyl moiety of oxazepam
L Kangas et al.
European journal of clinical pharmacology, 17(4), 301-304 (1980-04-01)
Six women, 13 to 16 weeks pregnant, and 12 women at 38 to 40 weeks gestation, received oral oxazepam about 12 h before legal abortion, by hysterotomy in the former and before elective caesarean section in the latter group. The
J Sonne et al.
European journal of clinical pharmacology, 35(4), 385-389 (1988-01-01)
Six healthy volunteers received oxazepam 15 mg i.v. and orally at an interval of at least one week. The kinetic variables of i.v. oxazepam were: elimination half-life (t1/2 beta) 6.7 h, total clearance (CL) 1.07 ml.min-1.kg-1, volume of distribution (Vc)

Protocols

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service