Skip to Content
Merck
All Photos(1)

Documents

L-020

Supelco

Levetiracetam solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14N2O2
CAS Number:
Molecular Weight:
170.21
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CC[C@H](N1CCCC1=O)C(N)=O

InChI

1S/C8H14N2O2/c1-2-6(8(9)12)10-5-3-4-7(10)11/h6H,2-5H2,1H3,(H2,9,12)/t6-/m0/s1

InChI key

HPHUVLMMVZITSG-LURJTMIESA-N

Gene Information

General description

A certified Snap-N-Spike® solution applicable for use in levetiracetam testing methods by LC/MS or GC/MS for applications in clinical toxicology, medication monitoring, urine drug testing, pharmaceutical research, or forensic analysis. Levetiracetam is an antiepileptic drug used in the treatment of epilepsy and is marketed under the trade name Keppra®.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Keppra is a registered trademark of UCB Pharma, S.A.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Consoli et al.
Cerebrovascular diseases (Basel, Switzerland), 34(4), 282-289 (2012-11-07)
Strokes are the leading cause of epileptic seizures in adults and account for 50% of seizures in those over the age of 65 years. The use of antiepileptic drugs to prevent recurrent poststroke seizures is recommended. One hundred and twenty-eight
Hai-Dun Yan et al.
Brain research bulletin, 90, 142-148 (2012-10-31)
Levetiracetam (LEV) is a widely used antiepileptic agent for partial refractory epilepsy in humans. LEV has unique antiepileptic effects in that it does not inhibit electroshock- or pentylenetetrazol-induced convulsion, but does inhibit seizures in kindling animal and spontaneously epileptic rat
Marjolein de Groot et al.
Neuro-oncology, 15(2), 216-223 (2012-12-13)
Treatment of high-grade glioma (HGG) patients with anti-epileptic drugs (AEDs) has met with various side effects, such as cognitive deterioration. The cognitive effects of both older and newer AEDs in HGG patients are largely unknown. The aim of this study
Radica M Stepanović-Petrović et al.
Anesthesia and analgesia, 115(6), 1457-1466 (2012-11-13)
We have recently shown that levetiracetam, administered systemically, exerts an antihyperalgesic effect in a rat inflammatory pain model. In this study, we examined whether levetiracetam has local peripheral antihyperalgesic/anti-edematous effects in the same model of localized inflammation and whether opioidergic
Justin B Usery et al.
Journal of neuro-oncology, 99(2), 251-260 (2010-02-11)
To determine the safety and tolerability of IV and oral levetiracetam monotherapy for seizures in brain tumor patients following resection. Brain tumor patients undergoing neurosurgery with >or=1 seizure within the preceding month prior to surgery were enrolled to receive intravenous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service