Skip to Content
Merck
All Photos(1)

Key Documents

D-023

Supelco

N-Desmethyl-cis-tramadol hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24ClNO2
CAS Number:
Molecular Weight:
285.81
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CNC[C@H]1CCCC[C@]1(O)C2=CC(OC)=CC=C2.[H]Cl

InChI

1S/C15H23NO2.ClH/c1-16-11-13-6-3-4-9-15(13,17)12-7-5-8-14(10-12)18-2;/h5,7-8,10,13,16-17H,3-4,6,9,11H2,1-2H3;1H/t13-,15+;/m1./s1

InChI key

NOZLWRHUQJHIRG-PBCQUBLHSA-N

General description

N-Desmethyl-cis-tramadol is a primary urinary and plasma metabolite of the synthetic analgesic tramadol. Tramadol, sold under trade names Ultram®, Conzip®, and Ryzolt®, relieves moderate to severe pain. This analytical reference standard is applicable for numerous LC/MS or GC/MS applications from pain prescription monitoring, urine drug testing and forensic analysis to clinical toxicology or pharmaceutical testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Conzip is a registered trademark of Cipher Pharmaceuticals Inc.
Ryzolt is a registered trademark of Purdue Pharma Products L.P.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Ultram is a registered trademark of Johnson & Johnson

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

This article presents an LC-MS/MS method for the determination of tramadol in urine samples using reversed-phase LC-MS/MS aided by isotope internal standards.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service