Skip to Content
Merck
All Photos(1)

Key Documents

880128P

Avanti

DSPE-PEG(2000) Amine

Avanti Research - A Croda Brand

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[amino(polyethylene glycol)-2000] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (880128P-10mg)
pkg of 1 × 100 mg (880128P-100mg)
pkg of 1 × 25 mg (880128P-25mg)
pkg of 1 × 50 mg (880128P-50mg)
pkg of 1 × 500 mg (880128P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOO/C=C\N)=O)=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.

General description

DSPE-PEG(2000) is a 18-carbon acyl chain containing phospholipid with a critical micelle concentration of 1× 106 M. It is conjugated with functional groups like biotin, primary amine and carboxylic acid. The micelles display spheroid shape with a size of 18mm.

Application

DSPE-PEG(2000) Amine has been used:
  • for the generation of single-walled carbon nanotubes (SWNTs) phospholipid-polyethylene glycol-folic acid conjugate
  • in the preparation of polyethylene glycol (PEGylated) magnetic nanoparticles based liposomes
  • in the preparation of liposomes for myomerger ectodomain based disruption studies

Biochem/physiol Actions

DSPE-PEG(2000) micelles aids in the solubilization of hydrophobic drugs and their delivery. It is used for coating of magnetic iron oxide-based nanoparticles.

Packaging

20 mL Clear Glass Screw Cap Vial (880128P-500mg)
5 mL Amber Glass Screw Cap Vial (880128P-100mg)
5 mL Amber Glass Screw Cap Vial (880128P-10mg)
5 mL Amber Glass Screw Cap Vial (880128P-25mg)
5 mL Amber Glass Screw Cap Vial (880128P-50mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functionalization and peptide-based delivery of magnetic nanoparticles as an intracellular MRI contrast agent
Nitin N, et al.
Journal of Biological Inorganic Chemistry, 9(6), 706-712 (2004)
Thermodynamic and kinetic stability of DSPE-PEG (2000) micelles in the presence of bovine serum albumin
Kastantin M, et al.
The Journal of Physical Chemistry B, 114(39), 12632-12640 (2010)
Myomaker and myomerger work independently to control distinct steps of membrane remodeling during myoblast fusion
Leikina E, et al.
Developmental Cell, 46(6), 767-780 (2018)
Enhanced blood-brain barrier transmigration using a novel transferrin embedded fluorescent magneto-liposome nanoformulation
Ding H, et al.
Nanotechnology, 25(5), 055101-055101 (2014)
The impact of subcellular location on the near infrared-mediated thermal ablation of cells by targeted carbon nanotubes
Murali VS, et al.
Nanotechnology, 27(42), 425102-425102 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service