Skip to Content
Merck
All Photos(1)

Key Documents

610003

Avanti

PC Membranes 0.05um

Avanti Research - A Croda Brand

Synonym(s):

Extruder PC Membrane, Polycarbonate Membranes 0.05μm 19mm (100/pk)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
NACRES:
NB.24

manufacturer/tradename

Avanti Research - A Croda Brand

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

PC Membranes possesses smooth and flat surfaces for filtration with of track-etched uniform cylindrical pores.
Polycarbonate Membrane for Avanti′s Mini-Extruder.
Watch the video to learn more about the Avanti Research Mini-Extruder.

Application

PC Membranes 0.05um has been used to extrude unilamellar liposomes.

Biochem/physiol Actions

PC Membrane exhibits fine mechanical properties, chemical stability, and non-auto fluorescent properties. It is widely used in immunofluorescent direct count method for the identification of various bacteria.

Components

Whatman® Nuclepore Hydrophilic Membrane 800308

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
Whatman is a registered trademark of Cytiva

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Formation of supported lipid bilayers containing phase-segregated domains and their interaction with gold nanoparticles
Melby ES, et al.
Environmental science. Nano, 3(1), 45-55 (2016)
Highly sensitive detection of Salmonella typhi using surface aminated polycarbonate membrane enhanced-ELISA
Jain S, et al.
Biosensors And Bioelectronics, 31(1), 37-43 (2012)
Determination of glycolipid-protein interaction specificity.
Lopez PHH and Schnaar RL
Methods in Enzymology, 417, 205-220 (2006)
Response of MG63 osteoblast-like cells onto polycarbonate membrane surfaces with different micropore sizes
Lee SJ, et al.
Biomaterials, 25(19), 4699-4707 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service