Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Z146714
Aldrich® non-corrosive lecture-bottle gas regulator
delivery pressure 0-22 psig, CGA Inlet size 170
About This Item
Recommended Products
material
stainless steel diaphragm
delivery pressure measuring range
0-22 psig
CGA Inlet size
170
General description
- Single-stage
- Inlet: CGA 170
- Outlet: male 1/4 NPTF
- Max. inlet pressure: 3,000 psig
- Temp. range: -40 to 140°F
- Compact chrome-plated brass body
- Lecture-bottle gas regulators include a PTFE sealing washer
- Order spare PTFE sealing washers separately
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
Already Own This Product?
Customers Also Viewed
-
What is the Department of Transportation shipping information for this product?
1 answer-
Helpful?
-
-
What is the product number for the PTFE washer for the Lecture Bottle Gas Regulators?
1 answer-
Lecture-bottle gas regulators include a PTFE sealing washer. Spare PTFE sealing washers can be ordered.Product Z146994, PTFE sealing washers, CGA Inlet size 180 can be used for Products Z148504, Aldrich® corrosive lecture-bottle gas regulator, 0-15 psig delivery pressure, CGA Inlet size 180 and Z146714, Aldrich® non-corrosive lecture-bottle gas regulator, 0-15 psig delivery pressure, CGA Inlet size 180.Product Z513105, PTFE sealing washers, CGA Inlet size 170 can be used for Products Z512958, Aldrich® non-corrosive lecture-bottle gas regulator, 0-15 psig delivery pressure, CGA Inlet size 170 and Z513113, Aldrich® non-corrosive lecture-bottle gas regulator, 0-100 psig delivery pressure, CGA Inlet size 170.
Helpful?
-