Skip to Content
Merck
All Photos(1)

Key Documents

W294705

Sigma-Aldrich

Propyl gallate

≥98%, FCC

Synonym(s):

3,4,5-Trihydroxybenzoic acid propyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3,4,5-(HO)3C6H2CO2CH2CH2CH3
CAS Number:
Molecular Weight:
212.20
FEMA Number:
2947
Beilstein:
1877976
EC Number:
MDL number:
UNSPSC Code:
12164502
eCl@ss:
39024506
PubChem Substance ID:
NACRES:
NA.21
Organoleptic:
odorless
grade:
Fragrance grade
Halal
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
EU Regulation 178/2002
FCC
FDA 21 CFR 172.615
FDA 21 CFR 175.125
FDA 21 CFR 175.300
FDA 21 CFR 175.380
FDA 21 CFR 175.390
FDA 21 CFR 184.1660

Assay

≥98%

mp

146-149 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

odorless

SMILES string

CCCOC(=O)c1cc(O)c(O)c(O)c1

InChI

1S/C10H12O5/c1-2-3-15-10(14)6-4-7(11)9(13)8(12)5-6/h4-5,11-13H,2-3H2,1H3

InChI key

ZTHYODDOHIVTJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propyl gallate is mainly used as an antioxidant for edible fats and vegetable oils. It occurs naturally in the pods of tara tree.

Reconstitution

Prepare a 0.1M solution in glycerol:PBS (9:1)

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

368.6 °F - closed cup

Flash Point(C)

187 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidation and antioxidants in fat and oil processing.
Sherwin ER.
Journal of the American Oil Chemists' Society, 55(11), 809-814 (1978)
Antioxidants for food fats and oils.
Sherwin ER.
Journal of the American Oil Chemists' Society, 49(8), 468-472 (1972)
A review on DNA interaction with synthetic phenolic food additives.
Dolatabadi JEN & Kashanian S.
Food Research International, 43(5), 1223-1230 (2010)
Lynn M Boyden et al.
The Journal of investigative dermatology, 135(6), 1540-1547 (2014-11-15)
Genetic investigation of inherited skin disorders has informed the understanding of skin self-renewal, differentiation, and barrier function. Erythrokeratodermia variabilis et progressiva (EKVP) is a rare, inherited skin disease that is characterized by transient figurate patches of erythema, localized or generalized
Tanvir Hussain et al.
European journal of medicinal chemistry, 44(11), 4654-4660 (2009-08-12)
A series of new 3-[4-(1H-imidazol-1-yl) phenyl]prop-2-en-1-ones were synthesized by the condensation of various acetophenones with 4-(1H-imidazol-1-yl) benzaldehyde which was itself prepared by the N-arylation of imidazole using hexadecyltrimethylammonium bromide as catalyst for the first time. All the synthesized compounds were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service