Skip to Content
Merck
All Photos(3)

Key Documents

W261114

Sigma-Aldrich

Lactic acid

greener alternative

natural, ≥85%

Synonym(s):

DL-Lactic acid, 2-Hydroxypropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)COOH
CAS Number:
Molecular Weight:
90.08
FEMA Number:
2611
Beilstein:
1209341
Council of Europe no.:
4c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.004
NACRES:
NA.21

grade

Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
FDA 21 CFR 117

Assay

≥85%

form

liquid

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.425 (lit.)

pH

2

bp

122 °C/15 mmHg (lit.)

density

1.209 g/mL at 25 °C (lit.)

cation traces

As: ≤3 ppm
Cd: ≤1 ppm
Fe: ≤10.0 ppm
Hg: ≤1 ppm
Pb: ≤10 ppm

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

odorless

SMILES string

CC(O)C(O)=O

InChI

1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)

InChI key

JVTAAEKCZFNVCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lactic acid is commonly used as a preservative in milk products and also as a flavoring agent.
Remaining 15% contains: water, higher oligomers of lactic acid and other FEMA GRAS components
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dictionary of Food Compounds with CD-ROM: Additives, Flavors, and Ingredients, 221-221 (2012)
Mohamed Ali Abdel-Rahman et al.
Biotechnology advances, 31(6), 877-902 (2013-04-30)
Fermentative production of optically pure lactic acid has roused interest among researchers in recent years due to its high potential for applications in a wide range of fields. More specifically, the sharp increase in manufacturing of biodegradable polylactic acid (PLA)
James Steele et al.
Current opinion in biotechnology, 24(2), 135-141 (2013-01-03)
It has been known since the 1960s that lactic acid bacteria are essential for the development of cheese flavor. In the ensuing 50 years significant research has been directed at understanding the microbiology, genetics and biochemistry of this process. This
Annabelle Olsson et al.
Journal of wildlife diseases, 49(3), 560-567 (2013-06-20)
Using a prospective, randomized study design we demonstrate that midazolam sedation minimizes acidosis compared with physical restraint in captive juvenile estuarine crocodiles during handling or noninvasive procedures at preferred body temperature. A dose of midazolam (5.0 mg/kg) was administered intramuscularly
Stephen Yiu Chuen Choi et al.
The Journal of pathology, 230(4), 350-355 (2013-06-05)
The common preference of cancers for lactic acid-generating metabolic energy pathways has led to proposals that their reprogrammed metabolism confers growth advantages such as decreased susceptibility to hypoxic stress. Recent observations, however, suggest that it generates a novel way for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service