Key Documents
SYNA341420
Synthware twin hose connecting adapter
joint: ST/NS 14/20
Synonym(s):
SynthwareA341420, Adapter, twin connecting hose
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A341420
Related Categories
General description
Adapter has two hose connections 180° from one another to permit a flow of inert gas over the apparatus.
Legal Information
Synthware is a trademark of Kemtech America Inc.