Skip to Content
Merck
All Photos(2)

Key Documents

M84607

Sigma-Aldrich

N-Methylthiourea

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CSNHCH3
CAS Number:
Molecular Weight:
90.15
Beilstein:
506162
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

crystals

mp

118-121 °C (lit.)

SMILES string

CNC(N)=S

InChI

1S/C2H6N2S/c1-4-2(3)5/h1H3,(H3,3,4,5)

InChI key

KQJQICVXLJTWQD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T S Plantinga et al.
Clinical pharmacology and therapeutics, 102(6), 1017-1024 (2017-05-10)
Agranulocytosis is a rare and serious adverse effect of antithyroid drugs, with unknown etiology. The present study aimed to uncover genetic susceptibility and underlying mechanisms of antithyroid drug-induced agranulocytosis (ATDAC). We studied two independent families with familial Graves' disease, of
Shruti Sharma et al.
International journal of molecular sciences, 14(1), 255-272 (2013-01-25)
Congenital heart defects with increased pulmonary blood flow (PBF) result in pulmonary endothelial dysfunction that is dependent, at least in part, on decreases in nitric oxide (NO) signaling. Utilizing a lamb model with left-to-right shunting of blood and increased PBF
S P Gorman et al.
The Journal of applied bacteriology, 60(4), 319-325 (1986-04-01)
The cidal activity of the antimicrobial agent, noxythiolin, was investigated against a laboratory strain and a fresh isolate of Candida albicans. The order of resistance to noxythiolin was hyphal form (isolate) greater than or equal to 25 degrees C-grown blastospores
[Proper use of noxythiolin: an experimental rat study on interaction with lactic acid].
J Maldonado et al.
Therapie, 35(3), 409-412 (1980-05-01)
S P Gorman et al.
Journal of clinical and hospital pharmacy, 10(2), 185-192 (1985-06-01)
The availability of a rapid and highly specific polarographic method of analysis for formaldehyde enabled investigation of the rate of formaldehyde release from noxythiolin solutions and actual concentrations of formaldehyde in solutions during clinical use and storage. The antimicrobial activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service