Skip to Content
Merck
All Photos(1)

Key Documents

M53701

Sigma-Aldrich

o-Methylisourea bisulfate

99%

Synonym(s):

2-Methylpseudourea monosulfate (1:1), OMI®

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC(OCH3)=NH · H2SO4
CAS Number:
Molecular Weight:
172.16
Beilstein:
3722928
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

118-120 °C (lit.)

SMILES string

COC(N)=N.OS(O)(=O)=O

InChI

1S/C2H6N2O.H2O4S/c1-5-2(3)4;1-5(2,3)4/h1H3,(H3,3,4);(H2,1,2,3,4)

InChI key

MDFRYRPNRLLJHT-UHFFFAOYSA-N

Legal Information

OMI is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheng Guo et al.
European journal of mass spectrometry (Chichester, England), 24(5), 384-396 (2018-07-26)
Modified peptides fragmented by collision-induced dissociation can offer additional sequence information, which is beneficial for the de novo sequencing of peptides. Here, the model peptide VQGESNDLK was carbamylated. The optimal conditions were as follows: temperature of 90℃, pH of 7
Dmitry S Loginov et al.
Parasites & vectors, 12(1), 212-212 (2019-05-08)
The availability of tick in vitro cell culture systems has facilitated many aspects of tick research, including proteomics. However, certain cell lines have shown a tissue-specific response to infection. Thus, a more thorough characterization of tick cell lines is necessary.
Jessica Z Bereszczak et al.
Journal of the American Society for Mass Spectrometry, 18(2), 201-207 (2006-10-28)
Development of a quantification method based on isotopic variants of O-methyl isourea (OMIU) in conjunction with reversed-phase (RP) liquid chromatography (LC) electrospray mass spectrometry is described for determining the relative quantification of tau-related peptides Ac-VQIVXK-NH2. Extracted ion chromatograms of the
V M Mahnir et al.
Toxicon : official journal of the International Society on Toxinology, 29(7), 819-826 (1991-01-01)
The effect of modification of amino groups on RTX-III induced lethality in mice has been studied. The toxicity was not affected by guanidination of one or two lysine residues with O-methylisourea, but guanidination of three or four lysine residues decreased
S I Shalabi et al.
The Journal of dairy research, 49(4), 607-617 (1982-11-01)
Several dicarbonyl compounds (glyoxal, substituted glyoxals, diacetyl and 1, 2-cyclohexanedione) had a marked stabilizing effect on the heat stability of milk, especially in the presence of urea. These reagents are believed to modify arginine more or less specifically suggesting an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service