Skip to Content
Merck
All Photos(2)

Key Documents

I0631

Sigma-Aldrich

Imidodiphosphate sodium salt

≥97%

Synonym(s):

Tetrasodium imidodiphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HNO6P2Na4
CAS Number:
Molecular Weight:
264.92
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97%

storage temp.

2-8°C

SMILES string

O=P([O-])([O-])NP([O-])([O-])=O.[Na+].[Na+].[Na+].[Na+]

InChI

1S/H5NO6P2.4Na/c2-8(3,4)1-9(5,6)7;;;;/h(H5,1,2,3,4,5,6,7);;;;/q;4*+1/p-4

InChI key

KDZOOFFPQVQSQG-UHFFFAOYSA-J

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N F Zakirova et al.
Bioorganicheskaia khimiia, 31(1), 96-102 (2005-03-25)
The preparation conditions for dichlorophosphinylphosphorimidic trichloride were optimized. It was used in the synthesis of esters of imidodiphosphoric acid. The interaction of the trichloride with amines resulted in the corresponding amidodiphosphates rather than in the expected amides of imidodiphosphoric acid.
T Rozovskaya et al.
FEBS letters, 247(2), 289-292 (1989-04-24)
It is demonstrated here that rat liver DNA polymerase beta catalyzes the pyrophosphorolysis reaction with pyrophosphate (PPi) and its analogues. The substrate specificity of the PPi-binding site of several DNA polymerases was investigated. It was discovered that the ability of
Anna M Rydzik et al.
Bioorganic & medicinal chemistry, 20(5), 1699-1710 (2012-02-10)
We describe synthesis and properties of eight dinucleotide mRNA 5' cap analogs containing imidodiphosphate moiety within 5',5'-tri- or tetraphosphate bridge (NH-analogs). The compounds were obtained by coupling an appropriate nucleoside 5'-imidodiphosphate with nucleotide P-imidazolide mediated by divalent metal chloride in
Highly efficient green and blue-green phosphorescent OLEDs based on iridium complexes with the tetraphenylimidodiphosphinate ligand.
Yu-Cheng Zhu et al.
Advanced materials (Deerfield Beach, Fla.), 23(35), 4041-4046 (2011-07-30)
A B Zyryanov et al.
Biochemistry. Biokhimiia, 70(8), 908-912 (2005-10-11)
Imidodiphosphate (the pyrophosphate analog containing a nitrogen atom in the bridge position instead of oxygen) is a potent inhibitor of family II pyrophosphatases from Streptococcus mutans and Streptococcus gordonii (inhibition constant Ki approximately 10 microM), which is slowly hydrolyzed by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service