Skip to Content
Merck
All Photos(1)

Key Documents

GF59253352

Polytetrafluoroethylene

(PTFE), sheet, thickness 1 mm, size 300 × 300 mm

Synonym(s):

FP303050

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352100
NACRES:
NA.23

form

sheet

manufacturer/tradename

Goodfellow 592-533-52

size

300 × 300 mm

thickness

1 mm

General description

For updated SDS information please visit www.goodfellow.com

Application


  • Design and Synthesis of P(AAm-co-NaAMPS)-Alginate-Xanthan Hydrogels and the Study of Their Mechanical and Rheological Properties in Artificial Vascular Graft Applications.: This study discusses the development of hydrogels for use in artificial vascular grafts, emphasizing the mechanical and rheological properties of the materials used, which include PTFE. (Li et al., 2024).

  • Synthesis, Optical Performance Characterization, and Durability of Electrospun PTFE-PEO Materials for Space Applications.: The study evaluates the durability and optical performance of PTFE-PEO materials, underlining their potential in harsh space environments. (Ibekwe et al., 2024).

  • Fabrication of Activated Carbon Derived from Glycerin Pitch for Desulfurization of Model Fuel Oil and Electrode Application.: This paper details the use of PTFE in the fabrication of electrodes for desulfurization and other industrial applications. (Zulkefli et al., 2024).


Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service