Key Documents
DWK883350-0575
Kimble® Disposable Pasteur Pipet
L 5.75 in., Borosilicate glass/PTFE
About This Item
UNSPSC Code:
41121500
NACRES:
NB.04
material
Borosilicate glass/PTFE
packaging
pkg of 1 packs
manufacturer/tradename
Kimble® 883350-0575
L
5.75 in.
General description
These Pasteur-type pipets are intended for one-time use in the transfer of small volumes.
Application
Bacteriology, immunology, hematology, and serology
Features and Benefits
Removes risk of cross-contamination of high value specimens and laboratory reagents.
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences