Skip to Content
Merck
All Photos(1)

Key Documents

D55792

Sigma-Aldrich

3,5-Dichloroaniline

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3NH2
CAS Number:
Molecular Weight:
162.02
Beilstein:
636492
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97%

form

crystals

bp

259-260 °C/741 mmHg (lit.)

mp

46-52 °C (lit.)

SMILES string

Nc1cc(Cl)cc(Cl)c1

InChI

1S/C6H5Cl2N/c7-4-1-5(8)3-6(9)2-4/h1-3H,9H2

InChI key

UQRLKWGPEVNVHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Target Organs

Kidney

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

271.4 °F - closed cup

Flash Point(C)

133 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Valentovic et al.
Toxicology letters, 78(3), 207-214 (1995-08-01)
3,5-Dichloroaniline (3,5-DCA), a derivative needed in the manufacture of dyes, pesticides and industrial compounds has been reported to induce renal damage. This study investigated whether pretreatment with inducers or inhibitors of P450 altered 3,5-DCA toxicity. P450 levels were induced in
C Lentza-Rizos et al.
Journal of agricultural and food chemistry, 48(6), 2522-2527 (2000-07-11)
Chlozolinate (Serinal) is a dicarboximide fungicide used in southern European countries principally on grapes. Maximum residue levels have not yet been set by FAO/WHO and are under evaluation in the EU. Field trials have been carried out in Greece on
Christian H Lindh et al.
Rapid communications in mass spectrometry : RCM, 21(4), 536-542 (2007-01-25)
The fungicides vinclozolin and iprodione are widely used in agriculture. These pesticides are dicarboximide fungicides containing the common moiety 3,5-dichloroaniline (3,5-DCA). It has been suggested that low-level exposures to such compounds may be associated with adverse health effects such as
A Vanni et al.
Chemosphere, 41(3), 453-458 (2000-11-01)
This paper describes the determination of 3,5-DCA in commercial composts, a common metabolite in a class of fungicides, and dicarboximides (Vinclozolin, Chlozolinate, Iprodione, Procymidone) which are commonly used in agriculture. The extracts, obtained in acetonitrile by sonication, are analysed by
Reversed-phase high-performance liquid chromatography of pesticides. VII. Analysis of Vinclozolin, Iprodione, Procymidone, Dichlozolinate and their degradation product 3,5-dichloroaniline on white must and wine extracts.
P Cabras et al.
Journal of chromatography, 256(1), 176-181 (1983-01-28)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service