Skip to Content
Merck
All Photos(1)

Key Documents

D207608

Sigma-Aldrich

N,N-Diphenylformamide

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2NCHO
CAS Number:
Molecular Weight:
197.23
Beilstein:
2209397
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

crystals

bp

337 °C/762 mmHg (lit.)

mp

69-73 °C (lit.)

SMILES string

[H]C(=O)N(c1ccccc1)c2ccccc2

InChI

1S/C13H11NO/c15-11-14(12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-11H

InChI key

DCNUQRBLZWSGAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne-Laure Gassner et al.
Science & justice : journal of the Forensic Science Society, 59(1), 58-66 (2019-01-19)
The present study aimed at providing data to assess the secondary transfer of organic gunshot residues (OGSR). Three scenarios were evaluated in controlled conditions, namely displacing a firearm from point A to point B, a simple handshake and an arrest

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service