Skip to Content
Merck
All Photos(1)

Documents

D161608

Sigma-Aldrich

N,N-Dimethylhydrazine

98%

Synonym(s):

1,1-Dimethylhydrazine, asym-Dimethylhydrazine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NNH2
CAS Number:
Molecular Weight:
60.10
Beilstein:
605261
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

1.94 (vs air)

vapor pressure

103 mmHg ( 20 °C)

Assay

98%

form

liquid

autoignition temp.

478 °F

expl. lim.

95 %

refractive index

n20/D 1.4075 (lit.)

bp

60-62 °C (lit.)

density

0.79 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CN(C)N

InChI

1S/C2H8N2/c1-4(2)3/h3H2,1-2H3

InChI key

RHUYHJGZWVXEHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N-Dimethylhydrazine can react with:
  • β-Naphthol via radical amination to form 1-amino-2-naphthol.
  • 2-Chloro- and 2,2-dichloro-(bromo)vinyl ketones via regioselective heterocyclization to form 3-substituted 1-methyl(5-halo)pyrazoles.
  • Terminal alkynes in the presence of TpRuCl(PPh3)2 (Tp = tris(pyrazolyl)borate) to form nitriles.


N,N-Dimethylhydrazine along with ferric chloride hexahydrate forms an effective reduction system for:
  • Synthesizing DNA binding pyrrolo[2,1-c][1,4]benzodiazepine (PBD) imines via reductive cyclization of the corresponding nitro aldehyde.
  • Transforming a variety of nitroarenes and azido compounds into the corresponding anilines and amino compounds, respectively.

Biochem/physiol Actions

Lung and colon tumor initiator in experimental animal model.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

14.0 °F - closed cup

Flash Point(C)

-10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Reaction of terminal alkynes with hydrazines to give nitriles, catalyzed by TpRuCl (PPh3)2: novel catalytic transformation involving a vinylidene ruthenium intermediate
Fukumoto Y, et al.
Organometallics, 21(19), 3845-3847 (2002)
Masaki Honda et al.
Marine drugs, 17(2) (2019-02-06)
The red macroalga Agarophyton chilensis is a well-known producer of eicosanoids such as hydroxyeicosatetraenoic acids, but the alga produces almost no prostaglandins, unlike the closely related A. vermiculophyllum. This indicates that the related two algae would have different enzyme systems
Guoguo Jin et al.
Oxidative medicine and cellular longevity, 2020, 7690845-7690845 (2020-06-23)
To explore fresh strategies in colorectal cancer (CRC) chemotherapy, we evaluated the capability of the ruthenium-phloretin complex in exterminating colon cancer by effectively addressing multiple apoptotic mechanisms on HT-29 cancer cells together with an animal model of colorectal cancer activated
On the reaction of β-naphthol with N, N-dimethylhydrazine-a new radical amination process
Barton DHR, et al.
Tetrahedron Letters, 24(15), 1601-1604 (1983)
An efficient synthesis of pyrrolo [2, 1-c][1, 4] benzodiazepine antibiotics via reductive cyclization
Kamal A, et al.
Bioorganic & Medicinal Chemistry Letters, 7(14), 1825-1828 (1997)

Articles

Cancer research has revealed that the classical model of carcinogenesis, a three step process consisting of initiation, promotion, and progression, is not complete.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service